eBioscience™ Essential Human Th1/Th17 Phenotyping Kit
Invitrogen™

eBioscience™ Essential Human Th1/Th17 Phenotyping Kit

The Invitrogen eBioscience Essential Human Th1/Th17 Phenotyping Kit is designed for flow cytometry-based identity testing of Type 1 and TypeRead more
Have Questions?
Catalog NumberQuantity
A42927100 reactions
Catalog number A42927
Price (USD)
1,524.00
Each
Quantity:
100 reactions
Price (USD)
1,524.00
Each
The Invitrogen eBioscience Essential Human Th1/Th17 Phenotyping Kit is designed for flow cytometry-based identity testing of Type 1 and Type 17 T–helper cells (Th1/Th17). It leverages high quality eBioscience antibodies, isotype controls, cell stimulation reagents (induces cytokine production), and fixation/permeabilization reagents (for intracellular markers IfN-g and IL-17) and includes a ready-to-use protocol for experimental setup, gating strategy, and analysis.

Benefits include:
• Proven performance―antibodies and supporting reagents designed for flow cytometry
• Highly referenced antibodies―includes one of the most referenced clones for IL-17
• Turnkey kit and protocol―contains reagents and standardized gating recommendations needed to perform flow cytometry characterization

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
FormatKit
Quantity100 reactions
Shipping ConditionApproved for shipment at Room Temperature or on Wet or Dry Ice
Product LineeBioscience
Product TypePhenotyping Kit
Unit SizeEach
Contents & Storage
  • CD4, Ifn-g, IL-17a antibodies, store at 2°C to 8°CC
  • Cell stimulating cocktail, store at -5 to -30°C
  • Protein transport cocktail, store at -5 to -30°C
  • Intracellular fixation & permealization reagents, store at 2°C to 8°CC
  • Isotype control, store at 2°C to 8°CC