Search
View additional product information for Ion 550™ Single Chip Supplemental Kit for Agrigenomics - FAQs (A43559)
2 product FAQs found
The Ion 550 Kit-Chef contains Ion Chef templating and sequencing consumables sufficient for preparing and sequencing eight Ion 550 chips for four Ion Chef runs and four sequencer initializations (dual chip run configuration). If you are running in the single chip configuration, then you will need to purchase the Ion 550 Single Chip Supplemental Kit separately to ensure that you have enough templating and sequencing consumables to support eight single chip Ion Chef templating runs and eight sequencer initializations with the Ion 550 Kit-Chef.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
The Ion 550 Single Chip Supplemental Kit contains pre-packaged templating supplies and sequencing reagent cartridges required to support eight single chip Ion Chef runs and eight sequencer initializations with the Ion 550 Kit-Chef. The Ion 550 Single Chip Supplemental Kit includes the following components:
- Ion S5 Chef Supplies (4 boxes) (ships at room temperature; store at room temperature)
- Ion S5 Sequencing Solutions (1 box) (ships at room temperature; store at room temperature)
- Ion S5 Sequencing Reagents (1box) (ships on wet ice at 2-8 degrees C; store at -20 degrees C)
IMPORTANT! Do not store the Ion S5 Sequencing Reagents (Part No. A27768) on dry ice or in a closed environment containing dry ice.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.