Attune™ NxT and CytPix Flow Cell Cleaning Solution - FAQs
Attune™ NxT and CytPix Flow Cell Cleaning Solution - FAQs
View additional product information for Attune™ NxT and CytPix Flow Cell Cleaning Solution - FAQs (A43635)
3 product FAQs found
Why do I have to use ultra pure water for dilution of the Attune NxT Flow Cell Cleaning Solution?
The Attune NxT Flow Cell Cleaning Solution is filtered to remove particulates to avoid scratching optical surfaces, it is important not to introduce particulates from the water, hence, we recommend using ultra pure water for dilution.
What is the pH of the Attune NxT Flow Cell Cleaning Solution?
The Attune NxT Flow Cell Cleaning Solution is at pH 12. This information is posted in the MSDS.
How do I use the Attune NxT Flow Cell Cleaning Solution?
The directions for use are provided on the vial label: Dilute 1:3 in ultra pure water. Use the solution with the SIP sanitize protocol just prior to each shutdown every day.