Product Image
Applied Biosystems™
TaqMan™ Liquid Biopsy dPCR Assay
TaqMan Liquid Biopsy digital PCR assays combine the TaqMan fluorogenic 5’ nuclease chemistry with digital PCR methodology to provide aWeitere Informationen
Have Questions?
KatalognummerMenge
A44177S (450 reactions), Made to Order
Katalognummer A44177
Preis (EUR)
1.592,65
Exklusiv online
1.790,00
Ersparnis 197,35 (11%)
Each
Menge:
S (450 reactions), Made to Order
TaqMan Liquid Biopsy digital PCR assays combine the TaqMan fluorogenic 5’ nuclease chemistry with digital PCR methodology to provide a sensitive, wet-lab validated solution for accurate detection of low-frequency mutant alleles in cancer research.
TaqMan Liquid Biopsy dPCR Assays have been optimized to work with digital PCR and droplet digital PCR systems to achieve the high performance required to detect and quantify mutations present at very low frequencies and have been wet-lab tested and are guaranteed to perform on the Applied Biosystems QuantStudio 3D and Bio-Rad QX-100 and QX-200 systems.
Overview of sensitive mutation detection using TaqMan Liquid Biopsy dPCR Assays:
• Optimized digital PCR performance—wet lab–verified TaqMan assays targeting over 100 common cancer mutations
• High sensitivity—detect and quantify rare mutant prevalence to ≤0.1%
• Cost-effective and convenient—single-tube format includes both wild type and mutant alleles
• Streamlined analysis—advanced algorithms for better quantification of rare mutations
• Guaranteed performance—assay replaced or account credited if you are not satisfiedif it doesn’t perform in accordance with our guarantee
• Custom design support—guidance available for Custom TaqMan SNP Genotyping Assays on the QuantStudio 3D Digital PCR System that are not part of this wet lab–verified collection.
How to order
TaqMan Liquid Biopsy dPCR assays can be added to your cart by using Quick Order and entering the Catalog Number and appropriate Assay ID.  Detailed information for each liquid biopsy dPCR assay, such as the Assay ID, nucleotide mutation, and context sequence can be found in the Assay Index File below.
All of our predesigned TaqMan Assays are covered by the TaqMan Assays qPCR Guarantee.*
*Subject to terms and conditionsTerms and conditions apply. For complete details, go to www.thermofisher.com/taqmanguarantee.
Specifications
Zur Verwendung mit (Geräte)QuantStudio™ 3D
ProduktlinieTaqMan
ProdukttypLiquid Biopsy dPCR Assay
MengeS (450 reactions), Made to Order
VersandbedingungRaumtemperatur
NachweisverfahrenPrimer-Sonde
Zur Verwendung mit (Anwendung)Genotypisierung
FormFlüssig
GC-Rich PCR PerformanceHoch
PCR-MethodeDigital PCR
Unit SizeEach
Inhalt und Lagerung
1 tube containing a 40X mix of pre-formulated assay (2 probes and 2 primers).
Store at -15° to -25°C.