Pair your qPCR Master Mix with ready to use TaqMan Gene Expression, Genotyping Assay or Custom Probes for multiplexing. Use our helpful TaqMan Search Wizard to find the Assays for your target of interest. Learn more
TaqMan™ Fast Advanced Master Mix for qPCR, no UNG
Applied Biosystems™

TaqMan™ Fast Advanced Master Mix for qPCR, no UNG

TaqMan Fast Advanced Master Mix delivers accurate quantification and dependability in a short time.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A443605 mL
A443591 mL
Catalog number A44360
Price (USD)
531.00
Special offer
Online exclusive
Ends: 11-Dec-2026
708.00
Save 177.00
Each
Quantity:
5 mL
Price (USD)
531.00
Special offer
Online exclusive
Ends: 11-Dec-2026
708.00
Save 177.00
Each

TaqMan Fast Advanced Master Mix delivers accurate quantification and dependability in a short time. It provides best-in-class performance, including the widest quantifiable range, in both single and duplex reactions, even with challenging targets. TaqMan Fast Advanced Master Mix contains AmpliTaq Fast DNA Polymerase, dNTPs with dUTP, ROX dye (passive reference), and optimized buffer components. It is supplied at a 2X concentration. Note: This TaqMan Fast Advanced Master Mix does not contain uracil-N-glycoslyase (UNG).

TaqMan Fast Advanced Master Mix features include:

  • Best-in-class performance—superior sensitivity, accuracy, dynamic range, and specificity compared to standard mixes in standard mode
  • Engineered for enhanced benchtop stability—stable at room temperature for up to 72 hours in preassembled reactions
  • Optimized for multiplexing—validated for duplexing with exogenous and endogenous internal positive control assays
  • Reduced run times (<40 minutes) on fast and standard instrumentation—optimized for fast mode on fast instruments and fast cycling conditions on standard instruments
  • Seamlessly transitions into your experiments—validated with TaqMan Assays for gene expression and microRNAs, and TaqMan array micro-fluidic cards

Best-in-class performance

TaqMan Fast Advanced Master Mix has been designed to provide performance equal to or better than standard master mixes. It has been benchmarked against the leading suppliers' standard and fast master mixes to help ensure that it succeeds in providing best-in-class sensitivity, accuracy, dynamic range, and specificity. TaqMan Fast Advanced Master Mix's dynamic range (up to 7 logs) is the widest in the industry. The impressive sensitivity of the mix is showcased in the figure below which compares the Ct values of TaqMan Fast Advanced Master Mix and TaqMan Universal PCR Master Mix across a panel of gene expression assays.

Benchtop stability for high-throughput handling and convenience

TaqMan Fast Advanced Master Mix has been engineered to retain a high level of performance in preassembled reactions for up to 72 hours. The stability of this mix provides users of high-throughput liquid handling systems the assurance that the results on the first plate will mimic those of the last plate. The figure below shows an assay that was run upon assembly (time 0) and after 72 hours of incubation at 30°C, simulating the most extreme room temperature scenario.

For those with less extreme throughput needs, the enhanced stability of this master mix provides an overall added convenience to your workflow, as you are no longer constrained to immediately running plates upon assembly.

Optimized for multiplexing

We realize that confidence is paramount when it comes to your results. TaqMan Fast Advanced Master Mix has been designed to help deliver accurate results for duplex reactions using an internal positive control (IPC). The figure below shows results for the experimental target β-actin (gene name: ACTB), which was serially diluted and amplified in single-target reactions and duplex reactions.

Validated for microRNA assays

TaqMan Fast Advanced Master Mix has been validated for use with multiple real-time PCR applications, including microRNA assays. The formulation provides high specificity and dynamic range, the two most critical performance attributes that define successful results when working with microRNAs. The data in the figure below demonstrate excellent PCR linearity over a 6-log range of input template.

Reduced run times on standard instrumentation

TaqMan Fast Advanced Master Mix has been optimized for use with both fast and standard instrumentation, enabling researchers who currently own standard instruments to also reap the performance benefits and time savings this mix provides. The figure below showcases the impressive results achieved when using TaqMan Fast Advanced Master Mix under fast thermal cycling conditions on the Applied Biosystems 7300 Real-Time PCR System. The mix has been tested with all Applied Biosystems standard real-time PCR instrumentation to enable success independent of whether or not you own a fast-enabled instrument.

NOTE: The product user guide can be downloaded below. When using this no-UNG version of TaqMan Fast Advanced Master Mix, the optional UNG incubation at 50°C for 2 minutes can be omitted.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Fidelity (vs. Taq)2 X
For Use With (Equipment)7500 Fast System, 7500 System, 7900HT System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 6 Pro, QuantStudio™ 7 Flex, QuantStudio™ 7 Pro, StepOne™, StepOnePlus™, ViiA™ 7 System, Applied Biosystems AmpQuest 96
IncludesdUTP
Inhibitor Tolerance (Simple)High
Multiplex CapabilityDuplex
No. of Reactions500 Reactions
Passive Reference DyeROX (Pre-mixed)
PolymeraseAmpliTaq Fast DNA Polymerase
Product LineTaqMan
Product TypeFast Advanced Master Mix
Quantity5 mL
Sample TypeDNA
Shipping ConditionWet Ice
Target SpecificitycDNA or gDNA only
Thermostability72 hr., Assembled Reaction
Concentration2X
Detection MethodPrimer-probe
For Use With (Application)Gene Expression, DNA Quantification, Presence/Absence
FormLiquid
GC-Rich PCR PerformanceHigh
PCR MethodqPCR
Reaction SpeedFast or Standard
Unit SizeEach
Contents & Storage
Contains 1 tube (5 mL) 2X TaqMan Fast Advanced Master Mix sufficient for 500 reactions

TaqMan Fast Advanced Master Mix contains AmpliTaq Fast DNA Polymerase, dNTPs (with dUTP), ROX dye (passive reference), and optimized buffer.

Store at 2°C to 8°C in the dark.

Frequently asked questions (FAQs)

I am using the TaqMan Gene Expression Cells-to-CT kit. Can I use the TaqMan Fast Advanced Master Mix in place of the TaqMan Gene Expression Master Mix to set up the qPCR reaction?

Yes, the TaqMan Fast Advanced Master Mix can be used in place of the TaqMan Gene Expression Master Mix when setting up the qPCR reaction for the TaqMan Gene Expression Cells-to-CT kit.