What precautions should I take while handling IonCode Barcode Adapters, 1153-1536?
When handling IonCode barcode adapters, be careful to avoid cross‑contamination by changing gloves frequently.
- Pulse vortex the IonCode Barcode Adapters plate several times.
- Centrifuge the IonCode Barcode Adapters plate for 1 min at 1000 x g to collect contents at the bottom of the wells.
- Carefully remove and then discard the plate seal.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
- Pulse vortex the IonCode Barcode Adapters plate several times.
- Centrifuge the IonCode Barcode Adapters plate for 1 min at 1000 x g to collect contents at the bottom of the wells.
- Carefully remove and then discard the plate seal.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.