MicroSEQ™ ID Microbial Identification Software v3.1.3, Lite, with Software Library Bundle - FAQs
MicroSEQ™ ID Microbial Identification Software v3.1.3, Lite, with Software Library Bundle - FAQs
View additional product information for MicroSEQ™ ID Microbial Identification Software v3.1.3, Lite, with Software Library Bundle - FAQs (A46530)
5 product FAQs found
What is included in the MicroSEQ ID Microbial Identification Software v3.1.3, Lite, with Software Library Bundle (Cat. No. A46530)?
The components of the MicroSEQ ID Microbial Identification Software v3.1.3, Lite, with Software Library Bundle (Cat. No. A46530) are the following:
- MicroSEQ ID Microbial Identification Software, v3.1.3, Lite, WIN 10
- MicroSEQ ID Microbial Identification Software, v3.1.3, Lite License
- MicroSEQ ID Fungal Gene Library, v2018
- MicroSEQ ID Fungal Gene Library, v2018 License
- MicroSEQ ID 16S rDNA 500 Microbial Library, v2019
- MicroSEQ ID 16S rDNA 500 Microbial Library, v2019 License
- MicroSEQ ID 16S rDNA Full Gene Library
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What operating system is MicroSEQ ID Microbial Identification Software, Lite, version 3.1.3 supported on?
MicroSEQ ID Microbial Identification Software, Lite, version 3.1.3 is supported on Microsoft Windows 10, 64-bit operating system.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What are the recommended computer specifications for the MicroSEQ ID Microbial Identification Software, Lite, version 3.1.3?
Here are the recommended computer specifications for the MicroSEQ ID Microbial Identification Software, Lite, version 3.1.3:
- Processor: Intel Core i5 processor, 2.8 GHz or faster
- Hard drive: ≥250 GB for software installation and data storage
- Memory: ≥16 GB
- Operating System (OS): Windows 10 IOT, 64‑bit
IMPORTANT! We strongly recommend that you use computers that are provided by Thermo Fisher Scientific. All validation and verification studies were performed using Thermo Fisher Scientific computers.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What are the new features of the MicroSEQ ID Microbial Identification Software, Lite, version 3.1.3?
No new features were introduced into MicroSEQ ID Microbial Identification Software, Lite, version 3.1.3, compared to version 3.1. MicroSEQ ID Microbial Identification Software, Lite, version 3.1.3 is a stand-alone version of MicroSEQ ID Microbial Identification Software, version 3.1.3.
The Lite and System edition of MicroSEQ ID Microbial Identification Software are now compatible with 64-bit Windows 10 operating systems.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What is the MicroSEQ ID Microbial Identification Software, Lite, version 3.1.3 used for?
The MicroSEQ ID Microbial Identification Software, Lite, version 3.1.3 allows you to easily identify and classify bacterial or fungal sequences by comparing them to a validated microbial library (available separately). This Lite version of the software is for use on a stand-alone PC (not connected to the genetic analyzer) for data analyses only.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.