GeneArt™ Gibson Assembly HiFi Master Mix
Invitrogen™
GeneArt™ Gibson Assembly HiFi Master Mix
GeneArt Gibson Assembly HiFi Master Mix enables DNA assembly and cloning via a technique that allows multiple overlapping DNA fragments to be seamlessly linked in a one-step, 15–60 minute isothermal reaction.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
A4662710 Reactions
A4662850 Reactions
A46629200 Reactions
카탈로그 번호 A46627
제품 가격(KRW)
278,000
Each
수량:
10 Reactions
제품 가격(KRW)
278,000
Each
GeneArt Gibson Assembly HiFi Master Mix enables seamless, one-step, single-tube assembly of multiple overlapping DNA fragments in an isothermal reaction. This flexible DNA assembly methodology works for both small and large constructs, accommodating single or multiple inserts. PCR amplified fragments can be assembled without purification and transformed following a rapid competent cell transformation protocol. This approach significantly reduces hands-on time without great losses in positive clone yields. The resulting products are suitable for various downstream applications, including competent cell transformation, PCR, rolling circle amplification (RCA), synthetic biology, in vitro cell-free workflows, and mutagenesis.
Features of the GeneArt Gibson Assembly HiFi Master Mix include:
  • One-step, single-tube reaction with scarless fragment assembly—simplifies the cloning process by using complementary overlaps to join DNA fragments without the need of restriction enzymes and ligases
  • High accuracy and fidelity—precise assembly with low rates of junction errors, providing high sequence fidelity
  • Multiple insert assembly for a wide range of fragment sizes—capable of assembling up to five DNA fragments plus a vector in one reaction, suitable for assembling DNA fragments ranging from 0.5 to 32 kb
  • Trusted and widely cited—over 4000 citations in the scientific literature, highlighting its great success and reliability
Seamless assembly of multiple fragments
GeneArt Gibson Assembly HiFi Master Mix, with its proprietary enzyme blend, is used to seamlessly assemble DNA fragments with shared terminal end homology without leaving any extra sequences behind. The required 20- to 40-base pair terminal end overlaps are designed into the de novo fragment for synthesis or can be easily engineered by PCR amplification with custom DNA oligos. After a 15–60 minute incubation, a portion of the assembly reaction is then transformed into chemically competent or electrocompetent cells. This special enzyme mix creates a seamless and covalently bound DNA construct with high efficiency.
Provides great versatility
GeneArt Gibson Assembly HiFi has emerged as a powerful molecular cloning method that enables researchers to engineer constructs that would be too cumbersome or impossible using traditional techniques. This versatile technique can be used in various genetic engineering, synthetic biology, and genome editing applications. Technical notes are provided for site-directed mutagenesis cell-free protein expression workflow combined with RCA.
Convenient formats
GeneArt Gibson Assembly HiFi kits are available in multiple formats, including:
  • GeneArt Gibson Assembly HiFi Cloning Kit, chemically competent cells (Cat. No. A46624 with One Shot TOP10 chemically competent cells)
  • GeneArt Gibson Assembly HiFi Cloning Kit, electrocompetent cells (Cat. No. A46626 with ElectroMAX DH10B electrocompetent cells)
  • GeneArt Gibson Assembly HiFi Master Mix kits (cloning kit without the cells) (this page)
For more challenging DNA assembly applications such as cloning large or up to 15 fragments simultaneously, GeneArt Gibson Assembly EX should be considered (Cat. No. A46633, A46634, A46635, A46636, A46637).
For Research Use Only. Not for use in diagnostic procedures.
사양
클로닝 방법Seamless cloning
제어 유형Positive Control
용도(애플리케이션)Seamless Cloning & Genetic Assembly
반응 수10 Reactions
단편 수Up to 6 fragments
제품라인GENEART
제품 유형HiFi Master Mix
수량10 Reactions
배송 조건Ambient, Wet Ice, Dry Ice
워크플로우 단계DNA Assembly
Unit SizeEach
구성 및 보관
Includes master mix, positive control, and water and accommodates the use of your own competent cells.
자주 묻는 질문(FAQ)
How big do DNA fragment homology arms need to be when using Gibson Assembly kits?
For fragments that are 8 kb or lesser in length, an overlap of 20-40 bp is recommended. For fragments greater than 8 kb in length, an overlap of 40-100 bp is recommended.
What type of vector can I use with Gibson Assembly kits?
Any linearized vector can be used with Gibson Assembly kits. We recommend using high-copy vectors for small inserts (<10 kb) and low-copy vectors for large inserts or toxic genes.
What type of inserts can I use with Gibson Assembly kits?
Any linearized DNA fragment, including PCR products, restriction digestion products, and GeneArt Strings can be used with Gibson Assembly kits.
What is the difference between the Gibson Assembly HiFi and the Gibson Assembly EX kits?
Gibson Assembly HiFi kits provide the quickest and easiest way to assemble up to six fragments in a single-tube reaction. The more robust Gibson Assembly EX kits enable assembly of up to 15 fragments in a single-tube, two-step reaction.