PureQuant CD3+ T Cell Assay
Applied Biosystems™

PureQuant CD3+ T Cell Assay

The Applied Biosystems PureQuant assays are a family of assays that accurately identify and quantify specific immune cell types inRead more
Have Questions?
Catalog NumberQuantity
A4719728 reactions
Catalog number A47197
Price (USD)
4,220.00
Each
Quantity:
28 reactions
Price (USD)
4,220.00
Each
The Applied Biosystems PureQuant assays are a family of assays that accurately identify and quantify specific immune cell types in a mixed population of cells. These assays are TaqMan qPCR-based and measure genomic DNA methylation at specific sites that serve as a unique identifier. The assays include standards and controls and each 96-well plate can accommodate up to seven test samples. Results are provided as a percentage of target cell type out of total input cells using a free analysis tool.

The PureQuant CD3+ Assay utilizes a specific demethylated site in the intragenic CD3G/CD3D region to positively identify CD3+ T-cells (see https://www.ncbi.nlm.nih.gov/pmc/articles/PMC3278789/).

Benefits include:
Proven performance―core technology has been used for identification and quantification of immune cell types for over a decade
Easily standardized―qPCR method enables consistent results across multiple users, instruments, and sites
Facilitates regulatory filings―established performance specifications following ICHQ2(R1) guidelines
Scheduling flexibility―assay can be performed using fresh or frozen cells/gDNA

The assays include all reagents, standards, and controls required to run the assay. Specific components include:
Calibrator: corrects for variation in bisulfite conversion efficiency. Plasmid-based calibrator harbors the genomic target regions in equimolar ratios and includes GAPDH. Subjected to bisulfite conversion followed by copy number estimation via qPCR standard curve. Allows estimation of assay-specific bisulfite efficiency factor: the ratio between TpG/GAPDH copy numbers measured from the same plasmid.
Reference: checks assay performance. Consists of genomic DNA extracted from human blood of pooled healthy donors. The reference values represent the expected range of the target cell type. For example, CD8+ cells are generally more abundant than Treg or Th17 cells.
Standards: determine the unknown copy number. Consist of six dilutions spanning 4-log linear dynamic range (30-31250 copies). Absolute copy numbers can reliably be determined using the slope and intercept of the respective standard curves.

This product is manufactured at a site with an ISO9001-certified quality management system.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)QuantStudio™
Format96-Well Plate
No. of Reactions28 reactions
Product TypePurequant CD3 + T Cell Assay
Quantity28 reactions
Sample TypeCell
Detection MethodPrimer-probe
FormFrozen
PCR MethodqPCR
Reaction SpeedFast or Standard
Unit SizeEach
Contents & Storage
•Dynabeads SILANE Genomic DNA Kit, store at 2°C to 8°C
• 1 x 2 mL PureQuant qPCR Master Mix II, store at 2°C to 8°C
• 4 x ABS
• THFA, store at room temperature
• PureQuant CD3 Module, store at -15° to -25°C (CD3 and GAPDH TaqMan primers and probes, Reference DNA, Lambda DNA, Standard DNA)