Pierce™ Yeast Digest Standard - FAQs
Pierce™ Yeast Digest Standard - FAQs
View additional product information for Pierce™ Yeast Digest Standard - FAQs (A47951)
1 product FAQs found
Which LC/MS analysis process does Yeast Digest Standard help monitor?
Yeast Digest Standard is suitable for monitoring the Ion Source parameters (spray stability, source temperature of ionization, in-source fragmentation) and Mass Analyzer components (mass accuracy from scan to scan) of the LC/MS analysis.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.