Pierce™ HeLa Digest/PRTC Standard, 10 μg - FAQs
Pierce™ HeLa Digest/PRTC Standard, 10 μg - FAQs
View additional product information for Pierce™ HeLa Digest/PRTC Standard - FAQs (A47997, A47996)
2 product FAQs found
What is the composition of HeLa Digest/PRTC Standard?
HeLa Digest/PRTC Standard is a combination of Pierce HeLa Protein Digest Standard (Cat. No. 88328 and 88329) and Pierce Peptide Retention Time Calibration Mixture (PRTC) (Cat. No. 88321). HeLa Digest/PRTC Standard contains 10 µg of HeLa Digest Standard and 5 pmol of PRTC.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
Which LC/MS analysis process does HeLa Digest/PRTC Standard help monitor?
HeLa Digest/PRTC Standard helps monitor all the steps of LC/MS analysis: LC, Ion Source, and Mass Analyzer. HeLa Digest monitors Ion Source and Mass Analyzer, while PRTC monitors the LC component of the LC/MS analysis process.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.