What is the composition of HeLa Digest/PRTC Standard?
HeLa Digest/PRTC Standard is a combination of Pierce HeLa Protein Digest Standard (Cat. No. 88328 and 88329) and Pierce Peptide Retention Time Calibration Mixture (PRTC) (Cat. No. 88321). HeLa Digest/PRTC Standard contains 10 µg of HeLa Digest Standard and 5 pmol of PRTC.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.