Viral Production Medium, 10 L - FAQs
Viral Production Medium, 10 L - FAQs
View additional product information for Viral Production Medium - FAQs (A4817901, A4817902, A4817903)
1 product FAQs found
Do I need to supplement the Viral Production Medium prior to use?
Yes, prior to using the Viral Production Medium, it should be supplemented with 4 mM GlutaMAX Supplement.