CTS™ Viral Production Cells 2.0 - FAQs
CTS™ Viral Production Cells 2.0 - FAQs
View additional product information for CTS™ Viral Production Cells 2.0 - FAQs (A48400)
5 product FAQs found
Can I use CTS Viral Production Cells (Cat. No. A53956) with CTS AAV MAX products?
CTS Viral Production Cells 2.0 are specifically designed to work with CTS AAV MAX products to provide maximal adeno-associated virus (AAV) titers and optimal performance. CTS Viral Production Cells can be used but we don't recommend doing this as it will reduce system performance.
What is the highest density that CTS Viral Production Cells 2.0 can be cultured at?
CTS Viral Production Cells 2.0 achieve maximum cell densities of >1.2 x 10E7 cells/mL in shake flask cultures.
What is the passage stability of CTS Viral Production Cells 2.0?
We recommend using CTS Viral Production Cells 2.0 between passages 3 to 25 for adeno-associated virus (AAV) production.
How long do CTS Viral Production Cells 2.0 require to recover post-thaw before transfection?
We recommend allowing freshly thawed CTS Viral Production Cells 2.0 to recover in culture for at least three passages prior to transfection.
Are CTS Viral Production Cells 2.0 a clonal cell line?
Yes, CTS Viral Production Cells 2.0 cells are a clonal cell line derived from single cell cloning of 293-F cells.