AccuSEQ™ Real-Time PCR Detection Software v3.1 - FAQs
AccuSEQ™ Real-Time PCR Detection Software v3.1 - FAQs
View additional product information for AccuSEQ™ Real-Time PCR Detection Software v3.1 - FAQs (A48509)
1 product FAQs found
When using the AccuSEQ Real-Time PCR Detection Software v3.1 (Cat. No. A48509), for the double signature function, do the users who will sign have to have different roles or could they also hold the same role?
With the AccuSEQ Real-Time PCR Detection Software v3.1 (Cat. No. A48509), the administrator can define whether 1 or 2 signatures are required to sign an experiment. However, it is not possible to specify that the signatures have to come from different roles. The administrator can only define how many signatures (up to 10) are required to sign an experiment.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.