Countess™ 3 Automated Cell Counter, 1 each - FAQs
View additional product information for Countess™ 3 Automated Cell Counter - FAQs (A49865, A50302, AMQAX2000, A49891, A50298, A49862)
4 product FAQs found
What cell counter do you offer that can count live and dead cells, measure cell viability, cell size, has high-resolution touch screen, and comes with reusable slides?
We suggest the Countess 3 and 3 FL Automated Cell Counters with starter packages; please see the product pages linked below:
Countess 3 FL Automated Cell Counter Starter Package + ABRC Extended Warranty (Cat. No. A50303)
Countess 3 FL Automated Cell Counter Starter Package + REX Extended Warranty (Cat. No. A50299)
Countess 3 FL Automated Cell Counter Starter Package (Cat. No. A49866)
Countess 3 Automated Cell Counter Starter Package + ABRC Extended Warranty (Cat. No. A50302)
Countess 3 Automated Cell Counter Starter Package + REX Extended Warranty (Cat. No. A50298)
Countess 3 Automated Cell Counter Starter Package (Cat. No A49865)
For more information about out Countess 3 Automated Cell Counters See the brochure linked below:
linkCountess 3 Automated Cell Counters: A counter that thinks like a cell biologist
Are there any recommended parameter settings on the Countess 3/3FL Automated Cell Counter (Cat. Nos. AMQAF2000, AMQAX2000) for specific cell lines?
No. The Countess 3/3 FL Automated Cell Counter (Cat. Nos. AMQAF2000, AMQAX2000) parameter settings, especially brightness, can vary between instruments due to slight differences in lamp output and other factors.
What can we do with the FCS file generated by the Countess 3/3 FL Automated Cell Counter (Cat. Nos. AMQAF2000, AMQAX2000)?
The Countess 3/3FL Automated Cell Counter (Cat. Nos. AMQAF2000, AMQAX2000) FCS files can be uploaded to most flow cytometry software that can accept FCS files, such as FlowJo or the Attune NxT software. Using the flow cytometry software, you may see the distribution of all cells and adjust/fine-tune parameter settings for only the cells of interest or analyze outliers.
Can we use any EVOS Light Cube with the Countess 3 FL Automated Cell Counter (Cat. No. AMQAF2000)?
With the exception of EVOS Light Cube, White (Cat no. AMEP4672) any of the EVOS Light Cubes for fluorescence detection can be used on the Countess 3 FL Automated Cell Counter (Cat. No. AMQAF2000).