TrueMark™ Xeno Control, kanamycin resistance
Applied Biosystems™

TrueMark™ Xeno Control, kanamycin resistance

TrueMark Xeno Control, Kanamycin Resistance, serves as an exogenous process control of a known concentration that is spiked into aRead more
Have Questions?
Catalog NumberQuantity
A503841000 μL
Catalog number A50384
Price (USD)
-
Quantity:
1000 μL
TrueMark Xeno Control, Kanamycin Resistance, serves as an exogenous process control of a known concentration that is spiked into a sample at specific stages throughout the qPCR process. Measuring the concentration of the spiked-in sequence allows for its use as a serial quality control, and it can be used to monitor recovery efficiency and to identify false negative results such as those that occur due to the presence of PCR inhibitors in molecular detection workflows. This is of particular importance when working with sample types that have a high level of inhibition.

We recommend using this spike-in control for pathogen detection panels containing assays that detect beta-lactamase sequences.

This product contains a kanamycin resistance sequence and can be used as an alternative to the TaqMan Universal DNA Spike-In Control (Cat. Nos. A39175 and A39176).

For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatTube
Product LineTrueMark
Quantity1000 μL
Shelf Life1 year
Shipping ConditionDry ice
Volume1000 μL
Concentration2 x 105 Copies/μL
Detection MethodPrimer-probe
Unit SizeEach
Contents & Storage
Store at -15°C to -25°C.