How long are the Genexus Dx Sequencing Kit consumables stable on the Genexus Dx instrument?
The Genexus Dx Sequencing Kit consumables are stable on the Genexus Dx instrument for up to two weeks after initialization, allowing flexibility to use the chip lanes according to sample throughput needs.