AccelerOme™ Label-Free MS Sample Prep Kit
Thermo Scientific™
AccelerOme™ Label-Free MS Sample Prep Kit
The Thermo Scientific AccelerOme Label-Free MS Sample Prep Kit provides ready-to-go reagents for efficient and robust proteomics mass spectrometry (MS)Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A509451 x 36 Reactions
A509441 x 16 Reactions
Catalog number A50945
Price (USD)
1,174.00
Each
Quantity:
1 x 36 Reactions
Price (USD)
1,174.00
Each
The Thermo Scientific AccelerOme Label-Free MS Sample Prep Kit provides ready-to-go reagents for efficient and robust proteomics mass spectrometry (MS) sample preparation for use with the AccelerOme Automated Sample Prep System.
Features of the Label-Free MS Sample Prep Kit include:
• Standardized—EasyPep kit chemistry designed for use with AccelerOme Automated Sample Prep System
• Convenient—complete set of ready-to-use proteomic sample prep and clean-up reagents for easy loading onto instrument
• Robust—reagents combined with AccelerOme ecosystem provide consistent, high-quality results with technical replicates with <5% CV
• Flexible—reagents verified on system using cells, plasma, proteins, and tissue samples for 1-–100 μg samples
• Time-saving—instrumentation and reagents perform the work enabling time away from tedious hands-on sample preparation
The AccelerOme Label-Free MS Sample Prep Kit contains pre-formulated buffers, MS-grade trypsin/Lys-C enzyme mix, and peptide clean up cartridges for the processing of samples from protein lysis and digestion through peptide clean-up. The single-use components are conveniently provided in labeled trays for direct placement on the AccelerOme system. This kit provides a complete reagent solution for automated sample preparation using proteins, plasma, cells, and tissue to produce high quality, MS-ready peptides.
The AccelerOme reagent portfolio is an integral part of the AccelerOme Automated system solution to provide robust proteomic MS sample preparation. The instrument hardware is built on a proven liquid handling format for reproducible sample processing and optimized on-line μSPE sample clean-up for excellent peptide recovery. Peptide quantitation post-cleanup is achieved through accurate UV spectrophotometric analysis to determine peptide concentrations prior to LC-MS analysis. The complete system (kits + reagents + software) was designed to provide high quality peptides while improving productivity with an easy-to-use automated system.
Specifications
Quantity1 x 36 Reactions
Detection MethodUV Absorption
Product LineThermo Scientific
Product TypeReagent Kit
Starting MaterialProtein Samples
Unit SizeEach
Frequently asked questions (FAQs)
Is the AccelerOme Label-Free MS Sample Prep Kit compatible with the BCA protein assay?
Yes, the buffer in the AccelerOme Label-Free MS Sample Prep Kit, in which samples are resuspended, is compatible with the BCA protein assay.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
Is the Lysis Buffer (Cat. No. A45735) compatible with the AccelerOme Label-Free MS Sample Prep Kit?
The Lysis Buffer (Cat. No. A45735) has been designed to be used with manual EasyPep MS Sample Prep Kits. The format of the Lysis Buffer container is not compatible with the AccelerOme platform and therefore it cannot be used with the AccelerOme Label-Free MS Sample Prep Kit.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
Can I use the samples obtained with the AccelerOme Label-Free MS Sample Prep Kit directly for mass spectrometry?
Yes, all buffers used in the AccelerOme Label-Free MS Sample Prep Kit are MS-compatible, and the samples can be directly used for mass spectrometry.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
Can the AccelerOme Label-Free MS Sample Prep Kit be reused?
The AccelerOme Label-Free MS Sample Prep Kit is a single-use kit, and it cannot be reused.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
Can I use the AccelerOme Label-Free MS Sample Prep Kit without the AccelerOme platform?
The AccelerOme Label-Free MS Sample Prep Kit has been developed to be used in the AccelerOme Automated Sample Preparation Platform. If you do not have the AccelerOme Automated Sample Preparation Platform, we recommend using EasyPep MS Sample Prep Kits (Cat. No. A40006, A57864, A45733 or A45734, depending on the number of reactions you would like to perform).
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.