Can I use the AccelerOme TMT11plex MS Sample Preparation Kits and the AccelerOme TMTpro 16plex MS Sample Preparation Kits for samples with protein amounts lower than 10 µg?
Unfortunately, the AccelerOme TMT11plex MS Sample Preparation Kits and the AccelerOme TMTpro 16plex MS Sample Preparation Kits have not been validated for use with sample lysates containing lower than 10 µg of initial protein.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.