AccelerOme™ TMTpro™ 16plex MS Sample Preparation Kits
Thermo Scientific™
AccelerOme™ TMTpro™ 16plex MS Sample Preparation Kits
TMTpro™ 16plex MS Sample Preparation Kits provide efficient and robust mass spec sample prep with TMTpro peptide labeling for use with the AccelerOme Automated Sample Prep System.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantitySufficient For
A509512 x 16 TMTpro Reactions30–100 μg samples
A509492 x 16 TMTpro Reactions10–30 μg samples
A509501 x 16 TMTpro Reactions30–100 μg samples
Catalog number A50951
Price (EUR)
3.913,00
Online exclusive
4.300,00
Save 387,00 (9%)
Each
Quantity:
2 x 16 TMTpro Reactions
Sufficient For:
30–100 μg samples
Price (EUR)
3.913,00
Online exclusive
4.300,00
Save 387,00 (9%)
Each
The AccelerOme TMTpro 16plex MS Sample Preparation Kits integrate pre-formulated buffers, MS-grade trypsin/Lys-C enzyme mix, and peptide clean-up cartridges with TMTpro 16plex label reagents for reproducible sample preparation of TMT-labeled peptides. The TMTpro 16plex label reagents have been optimized for use with samples from 10 to 100 μg samples to achieve greater than 95% labeling efficiency.
The Thermo Scientific Accelerome TMTpro 16plex MS Sample Preparation Kits provide a complete solution for efficient and robust proteomics mass spectrometry (MS) sample preparation, including TMT peptide labeling, for use with the AccelerOme Automated Sample Prep System.
TMTpro 16plex MS Sample Preparation Kits features:
• Flexible—sample prep and TMT-labeling reagents verified on system using cells, plasma, proteins, and tissue samples for 10 to 100 μg of sample input (kit dependent)
• Multiplex—enables concurrent MS analysis of up to 16 samples
• Optimized — EasyPep kit chemistry combined with TMTpro 16plex labeling reagents specifically designed for use with the AccelerOme Automated Sample Prep System
• Convenient—complete set of ready-to-use proteomic sample prep, clean-up, and TMT-labeling reagents for easy loading onto the instrument
• Robust —reagents combined with AccelerOme ecosystem provide reproducible, high-quality TMT-labeled peptides with greater than 95% labeling efficiency
Kit components are single-use and conveniently provided in labeled trays for direct placement on the AccelerOme system. This kit provides a complete reagent solution for automated sample preparation of TMTpro 16plex labeled samples to produce high quality, MS-ready peptides.
The AccelerOme reagent portfolio is an integral part of the AccelerOme Automated system solution to provide robust proteomic mass spec sample preparation. The instrument hardware is built on a proven liquid handling format for reproducible sample processing and optimized on-line μSPE sample clean-up for excellent peptide recovery. TMT label reconstitution and sample labeling and pooling have been incorporated before sample clean-up. Peptide quantitation post-clean-up is achieved through accurate UV spectrophotometric analysis to determine peptide concentrations prior to LC-MS analysis. The complete system (kits + reagents + software) was designed to provide high quality peptides while improving productivity with an easy-to-use automated system. When combined with the industry-leading, high-resolution Orbitrap instruments and software, TMTpro 16plex reagents provide integrated total solutions for quantitative protein expression analysis.
These products are subject to a limited use label license.
Specifications
Final Product TypeTMT-labeled peptide sample preparation
Quantity2 x 16 TMTpro Reactions
Sufficient For30–100 μg samples
Workflow StepAutomated mass spec sample prep
Detection MethodUV Absorption
FormatKit
Product LineThermo Scientific
Product TypeReagent Kit
Starting MaterialProtein Samples
Unit SizeEach
Contents & Storage
Sample prep module, TMT module
Frequently asked questions (FAQs)
Can I use the AccelerOme TMT11plex MS Sample Preparation Kits and the AccelerOme TMTpro 16plex MS Sample Preparation Kits for samples with protein amounts lower than 10 µg?
Unfortunately, the AccelerOme TMT11plex MS Sample Preparation Kits and the AccelerOme TMTpro 16plex MS Sample Preparation Kits have not been validated for use with sample lysates containing lower than 10 µg of initial protein.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
With the AccelerOme TMT11plex MS Sample Preparation Kits and the AccelerOme TMTpro 16plex MS Sample Preparation Kits, can I use any other lysis buffer instead of the one in the kit?
We do not recommend using a different lysis buffer other than the one included in the kit. On the one hand, variation in the lysis buffer might affect the subsequent sample prep steps. On the other hand, volume of the different buffers included in the kit has been optimized for the correct performance of the Accelerome platform.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
When using the AccelerOme TMT11plex MS Sample Preparation Kits and the AccelerOme TMTpro 16plex MS Sample Preparation Kits with the AccelerOme platform, can I stop the process at any time?
We do not recommend stopping the automatized process as the AccelerOme platform is programmed to perform the complete sample preparation in a single run while keeping sample stability and reproducibility as its highest.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
When using the AccelerOme TMT11plex MS Sample Preparation Kits and the AccelerOme TMTpro 16plex MS Sample Preparation Kits, should I add a protease inhibitor cocktail to the lysis buffer? How about phosphatase inhibitors?
We do not recommend using protease inhibitor cocktails containing EDTA with the AccelerOme TMT11plex MS Sample Preparation Kits and the AccelerOme TMTpro 16plex MS Sample Preparation Kits as they will inhibit both Universal nuclease and protease Trypsin/Lys-C activities. Following cell lysis, reduction and alkylation, the Trypsin/Lys-C proteases mix is added for complete digestion. Therefore, if protease inhibitors containing EDTA are added, there will be incomplete proteolysis. On the other hand, we do recommend using a phosphatase inhibitor cocktail (e.g., Halt Phosphatase Inhibitor Cocktail) if a subsequent phosphopeptide enrichment analysis will be performed.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
Can I run the samples obtained from the AccelerOme TMT11plex MS Sample Preparation Kits and the AccelerOme TMTpro 16plex MS Sample Preparation Kits in any other mass spectrometer besides Orbitrap Mass Spectrometers?
We recommend using Orbitrap Mass Spectrometers as the analysis of TMT reagents requires a system that is able to perform high-resolution MS/MS fragmentation. Orbitrap Mass Spectrometers include a specific TMT MS2 workflow which enables unique acquisition management resulting in confident results without compromising coverage.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
Citations & References (1)
Citations & References
Abstract
Development of an Adaptive, Economical, and Easy-to-Use SP3-TMT Automated Sample Preparation Workflow for Quantitative Proteomics.
Authors:Hermanson JN,Barny LA,Plate L
Journal:bioRxiv : the preprint server for biology
PubMed ID:40060590
Liquid handling robots have been developed to automate various steps of the bottom-up proteomics workflow, however, protocols for the generation of isobarically labeled peptides remain limited. Existing methods often require costly specialty devices and are constrained by fixed workflows. To address this, we developed a cost-effective, flexible, automated sample preparation ... More