T. foetus Reagents TAMUC

T. foetus Reagents TAMUC

Applied Biosystems T. foetus Reagents TAMUC are for the amplification of Tritrichomonas foetus RNA & DNA from smegma samples collectedRead more
Have Questions?
Catalog NumberQuantity
A51086100 reactions
Catalog number A51086
Price (USD)
600.00
Each
Quantity:
100 reactions
Price (USD)
600.00
Each
Applied Biosystems T. foetus Reagents TAMUC are for the amplification of Tritrichomonas foetus RNA & DNA from smegma samples collected in PBS.

Features of the reagents include:
• The first commercial reverse transcription qPCR for amplification of Tritrichomonas foetus RNA & DNA
• Highly sensitive, qualitative real-time PCR
• Optimized multiplex PCR amplification
• Can be used to test pools of up to five samples

The amplification reaction consists of a single-well (or single-tube) RT-qPCR in which T. foetus RNA/DNA and Xeno Control RNA targets are amplified and detected in real time using fluorescent TaqMan probes. The reagents include:
• T. foetus/Xeno Primers Probes Mix—for optimized multiplex RT-qPCR amplification of T. foetus RNA/DNA and Xeno control RNA
• Fast Virus Master Mix
• Nuclease-free Water

Specifications
Kit TypeCustom reagents
No. of Reactions100 reactions
Quantity100 reactions
Sample TypeBull Smegma Sample
SpeciesBovine
Starting MaterialSmegma
Sufficient For100 Samples
TargetITS1
Target Organism ClassParasite
TechniqueRT-qPCR
For Use With (Application)Real Time PCR (qPCR)
Unit SizeEach