T. foetus Controls TAMUC

T. foetus Controls TAMUC

Applied Biosystems T. foetus Controls TAMUC (Trichomonas foetus/Xeno RNA and Xeno RNA Controls) serve as amplification controls in a RT-qPCR.Read more
Have Questions?
Catalog NumberQuantity
A51087100 reactions
Catalog number A51087
Price (USD)
600.00
Each
Quantity:
100 reactions
Price (USD)
600.00
Each
Applied Biosystems T. foetus Controls TAMUC (Trichomonas foetus/Xeno RNA and Xeno RNA Controls) serve as amplification controls in a RT-qPCR.

The controls include:
• T. foetus-Xeno Control RNA Mix
• Xeno Control RNA—serves as internal control for the RNA/DNA isolation process and as a monitor for the presence of PCR inhibitors

Specifications
Amplicon Size109 bp
DiseaseBovine Trichomoniasis
Elution Volume Max90 μL
Purification MethodMagnetic Beads
Purification Time30 min.
Quantity100 reactions
Unit SizeEach