Is VitroEase Apoferritin Standard suitable for high resolution cryo-EM (electron microscopy)?
VitroEase Apoferritin Standard is suitable for high resolution cryo-EM (electron microscopy) and we have imaged it at 1.8 angstroms using our Krios G4 Cryo-TEM instrument.