What are the shipping and storage conditions for the VitroEase Training Kit?
The VitroEase Training Kit comes with two modules that are shipped separately. The Apoferritin Standard is shipped on dry ice and should be stored at -80 degrees C. It is also sold separately as Cat. No. A51362. The accessories (grids, tweezers, and Cryo TEM Grid Box) are shipped and stored at room temperature.