Pierce™ Small Molecule System Suitability Standard - FAQs
Pierce™ Small Molecule System Suitability Standard - FAQs
View additional product information for Pierce™ Small Molecule System Suitability Standard - FAQs (A51740)
3 product FAQs found
In which mode can I run the Small Molecule Suitability Standard?
The Small Molecule Suitability Standard can be run in Positive or Negative mode.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
Does the Small Molecule Suitability Standard need to be diluted?
The Small Molecule Suitability Standard can be diluted 1:10 (25 µL of standard with 225 µL of 0.1% Formic Acid) or can be used without dilution. Glass vials are provided for dilutions. It is important to use 0.1% Formic Acid solution for dilution, in order to keep all components in solution.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
What is the mass range for the Small Molecule Suitability Standard?
The mass range for the Small Molecule Suitability Standard (SMSS) is 76 to 1279 m/z.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.