Search
View additional product information for Absolute Q™ DNA Digital PCR Master Mix (5X) - FAQs (A52490)
1 product FAQs found
Yes, ROX is the passive reference dye for QC (Quality Control).
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.