Notice: It is STRONGLY recommended to read the user and/or quick reference guide below, especially when utilizing this mix in combination with pan-microbial assay(s).
Pair your qPCR Master Mix with ready to use TaqMan Gene Expression, Genotyping Assay or Custom Probes for multiplexing. Use our helpful TaqMan Search Wizard to find the Assays for your target of interest. Learn more
TaqPath™ BactoPure™ Microbial Detection Master Mix, no Rox
Applied Biosystems™

TaqPath™ BactoPure™ Microbial Detection Master Mix, no Rox

TaqPath BactoPure Microbial Detection Master Mix is optimized for rapid low-level microbial detection even in the presence of PCR inhibitors.Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume
A527041 x 5 mL
A527031 x 1 mL
A527055 x 1 mL
A527061 x 50 mL
Catalog number A52704
Price (USD)
913.65
Online exclusive
962.00
Save 48.35
Each
Volume:
1 x 5 mL
Price (USD)
913.65
Online exclusive
962.00
Save 48.35
Each

TaqPath BactoPure Microbial Detection Master Mix is optimized for rapid low-level microbial detection even in the presence of PCR inhibitors. All lots are functionally tested to help ensure lot-to-lot reproducibility for Ct and dynamic range consistency across a wide variety of assays. This General Purpose Reagent, labeled 'For Laboratory Use', is manufactured in an ISO 13485-certified and FDA-registered facility that adheres to the principles of current good manufacturing practices (cGMP).

Request an evaluation sample ›

Benefits of TaqPath BactoPure Microbial Detection Master Mix

  • Consistent low-level detection of bacterial, fungal, mammalian, and viral DNA, as well as antibiotic resistance markers
  • Tolerant of PCR inhibitors typically found in biopharmaceutical, molecular diagnostic, and research applications from both purified samples and crude lysates
  • Wide linear dynamic range* (up to 8 logs) enabling accurate detection with both low- and high-DNA-concentration samples
  • Optimized for multiplexing—available in two formulations that enable one- to four-target detection per reaction
  • 72 pre-PCR benchtop stability of pre-assembled** reactions, enabled by our proprietary hot-start mechanism
  • Compliance package containing the most frequently requested documents, available below

Two formulations

The master mix is available in two formulations:

  • TaqPath Bactopure Microbial Detection Master Mix containing ROX dye as a passive reference is recommended for singleplex (1 probe) and up to triplex (3 probes) reactions
  • TaqPath Bactopure Microbial Detection Master Mix (No ROX), which does not contain a passive reference, is recommended for use with assays utilizing the JUN (or similar omission wavelength) dye or for higher order multiplexing (>3 targets)

Consistent low-level detection

TaqPath BactoPure Microbial Detection Master Mix has been optimized for highly reproducible, low level detection from a wide variety of samples. While other commercially available mixes contain DNA background, which can lead to ambiguous results (figure below), each lot of TaqPath BactoPure master mix is tested on a panel of assays to certify that it is virtually free of such background prior to release. This enables TaqPath BactoPure master mix to reliably detect and quantitate low-concentration samples, as evident in the figure below. Also shown in a figure below is the consistent Cq results obtained from three unique lots across multiple assays, to maximize confidence in your results and highlight TaqPath BactoPure master mix's exceptional lot-to-lot consistency.

Tolerant of PCR inhibitors

The unique proprietary TaqPath BactoPure master mix formulation allows robust performance even in the presence of substances that can inhibit PCR (figure below), resulting in effective template detection from both purified DNA and crude lysate as illustrated in figure below.

Wide linear dynamic range

TaqPath BactoPure Microbial Detection Master Mix has been validated to provide a wide linear dynamic range* (up to 8 logs), enabling accurate detection with both low- and high-DNA-concentration samples. The formulation can be run in either fast or standard cycling conditions on a wide variety of qPCR platforms. The figure below demonstrates the excellent PCR linearity over an 8-log input range with a pan bacterial assay.

Optimized for multiplexing

TaqPath BactoPure Microbial Detection Master Mix has been validated for multiplexing up to four targets in one reaction, allowing for additional assays and/or controls to be run simultaneously for increased efficiency. Figure below illustrates this multiplexing capability by comparing performance between singleplex and multiplex reactions.

General purpose reagent labeled 'For Laboratory Use' and manufactured under cGMP

TaqPath master mixes are general purpose reagents labeled 'For Laboratory Use' and manufactured in an ISO 13485-certified and FDA-registered facility that adheres to cGMP principles.

Compliance package

In an effort to efficiently support your compliance needs, the following frequently requested documents have been made available below.

Not finding the compliance document you're looking for? Technical Support is here to help.

Learn more about multiplexing, TaqPath probes, and how our products can help with your projects ›

* Dynamic range is a function of the assay and template concentration in the sample, as well as the formulation of the master mix; thus, individual results may vary.

** Pre-PCR stability is influenced not only by the master mix, but also by the target being analyzed. For maximum confidence, stability profiles of specific targets should be confirmed.

For Laboratory Use
Specifications
For Use With (Equipment)7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, QuantStudio 12k Flex Dx, QuantStudio 12k Flex, QuantStudio 3, QuantStudio 5, QuantStudio 5 Dx, QuantStudio 6 Pro, QuantStudio 6 Flex, QuantStudio 7 Flex, QuantStudio 7 Pro, QuantStudio 7 Pro Dx, ViiA 7 System, ViiA 7 Dx, QuantStudio Dx, StepOnePlus System, 7500 Fast Dx System, StepOne System, Roche LightCycler PRO System, Bio-Rad CFX Opus 96 Real-Time PCR System
IncludesdUTP, UNG/UDG
Inhibitor Tolerance (Simple)Very High
Multiplex CapabilityUp to 4-plex
Passive Reference DyeNone
Product LineTaqPath
Quantity1 x 5 mL
Sample TypeDNA
SensitivityDown to 1 copy
Sufficient For500 x 20 μL Reactions
Target SpecificitycDNA or gDNA only
Thermostability72 hr., Assembled Reaction
Volume1 x 5 mL
Detection MethodPrimer-probe
For Use With (Application)Microbial Detection
PCR MethodqPCR
Reaction SpeedFast or Standard
Unit SizeEach
Contents & Storage
Contains 2X TaqPath BactoPure Microbial Detection Master Mix that includes:

• Hot-start AmpliTaq Fast DNA Polymerase
• Heat-labile uracil-DNA glycosylase (UNG)
• dNTPs (with dUTP)
• Optimized buffer

Store at 2–8°C.

Frequently asked questions (FAQs)

How do I download the Pathogen Interpretive Software?

Once you place your order, please contact our Instrument Service team at 1-800-955-6288 option 3, 1 (this kit is only available in North America). You will be given an access code to the TaqPath Monkeypox/Orthopox Virus DNA Kit eLearning where you’ll learn how to install, update, and download the Pathogen Interpretive Software package for the TaqPath Monkeypox/Orthopox Virus DNA Kit.

With the TaqPath Monkeypox/Orthopox Virus DNA Kit, what software should I use for data analysis?

Results are analyzed using the following software:
- Applied Biosystems Pathogen Interpretive Software v1.1
- SAE Administrator Console Dx v1.0 or v1.2 (for security and audit functions)
- Pathogen Interpretive Software 1.0.0 DAT (SAE Profile)

Please use the appropriate assay panel and template for your instrument:
- MPXOPX-EUA_QS5-9601_1.0.0.zip
- MPXOPX-EUA_QS5Dx-9602_1.0.0.zip
- MPXOPX-EUA_QS7F-384_1.0.0.zip

Note: These zip files are located in EducationConnect (www.thermofisher.com/educationconnect). The eLearning course needs to be completed to be granted access.

What is the instrument software configuration required to use the TaqPath Monkeypox/Orthopox Virus DNA Kit?

To obtain the correct analysis software or firmware version for your real-time PCR instrument, go to thermofisher.com/validatedfirmware, then select your instrument in the Real-Time PCR section.

Which instruments are authorized under the EUA for use with the TaqPath Monkeypox/Orthopox Virus DNA Kit?

Here are the instruments that can be used with the TaqPath Monkeypox/Orthopox Virus DNA Kit:
- Applied Biosystems QuantStudio 5 Real-Time PCR System (96-well, 0.1 mL block)
- Applied Biosystems QuantStudio 5 Dx Real-Time PCR System (96-well, 0.2 mL block)
- Applied Biosystems QuantStudio 7 Flex Real-Time PCR System (384-well block)

I would like to use the TaqPath Monkeypox/Orthopox Virus DNA Kit. Which extraction kit should I use to extract DNA from the sample?

Performance was established using the MagMAX Viral/Pathogen II Nucleic Acid Isolation Kit (Cat. No. A48383) in combination with the KingFisher Flex Magnetic Particle Processor with 96 Deep-Well Head and the KingFisher Flex 96 Deep-Well Heating Block (MVP_2Wash_400_Flex program). Other purification methods may lead to deviations in the expected result.