VetMAX™ PRRSV 3.0 Controls
Applied Biosystems™

VetMAX™ PRRSV 3.0 Controls

VetMAX™ PRRSV 3.0 Controls are for use in multiplex, real-time reverse transcription PCR (RT-PCR) assays targeting North American and European porcine reproductive and respiratory syndrome virus (PRRSV) and Xeno RNA.
Have Questions?
Catalog NumberQuantity
A5317010 Reactions
Catalog number A53170
Price (USD)
170.00
Each
Quantity:
10 Reactions
Price (USD)
170.00
Each

VetMAX PRRSV 3.0 Controls are for use in multiplex, real-time reverse transcription PCR (RT-PCR) assays targeting North American and European porcine reproductive and respiratory syndrome virus (PRRSV) and Xeno RNA. The controls come pre-mixed in one tube as a ready-to-use solution.

The included Xeno RNA Control is an internal positive control added to the lysis solution during RNA isolation to distinguish PCR inhibition, reaction failure, or human error from true negatives.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
DiseasePorcine Reproductive and Respiratory Syndrome Virus
No. of Reactions10 Reactions
Quantity10 Reactions
Shelf Life24 Months
Shipping ConditionShipped at -30°C to -10°C
Concentration2.5X
Unit SizeEach
Contents & Storage
Store at -10° to -30°C.