IN STOCK - Learn more about financing options and programs » Learn more
QuantStudio™ Absolute Q™ Digital PCR System, desktop, extended warranty, Smart Start
QuantStudio™ Absolute Q™ Digital PCR System, desktop, extended warranty, Smart Start
QuantStudio™ Absolute Q™ Digital PCR System, desktop, extended warranty, Smart Start
Applied Biosystems™
QuantStudio™ Absolute Q™ Digital PCR System, desktop, extended warranty, Smart Start
The Applied Biosystems QuantStudio Absolute Q Digital PCR System combines the power of digital PCR (dPCR) absolute quantification with theRead more
Have Questions?
Catalog NumberQuantity
A53267System + SmartStart + 1 yr ext warranty
Catalog number A53267
Price (USD)
-
Quantity:
System + SmartStart + 1 yr ext warranty
The Applied Biosystems QuantStudio Absolute Q Digital PCR System combines the power of digital PCR (dPCR) absolute quantification with the simplicity of a qPCR workflow. With a single hands-on step that takes five minutes to complete, dPCR is now easier than ever. The QuantStudio Absolute Q system provides a high quality, consistent, and easy-to-use solution for researchers who want to join the quantification revolution.
This system package includes an extended warranty and SmartStart orientation.
Features of the QuantStudio Absolute Q system include:
  • Easy and convenient qPCR-like workflow with only five minutes of hands-on time
  • Reduced errors and manual inputs with all functions integrated into a single instrument
  • Increased productivity with results in as little as 90 minutes
  • Reduced downtime with Smart Remote Support; show issues to Technical Support for speedy resolution
  • QuantStudio Absolute Q Digital PCR software enables intuitive setup, monitoring, and analysis
  • Security, auditing, and e-signature features for 21CFR part 11-compliance support
Easy and convenient workflow
The QuantStudio Absolute Q system was designed for a simple and powerful dPCR experience. All the necessary steps for dPCR—compartmentalization, thermal cycling, and data acquisition—are integrated into a single instrument. The simple qPCR-like workflow can be completed with under five minutes of hands-on time.
Accomplish more, faster
The QuantStudio Absolute Q system can complete a run in less than 90 minutes.
More information (including application notes)
QuantStudio Absolute Q assays
Get up and running fast with SmartStart Orientation
The included SmartStart Orientation is performed by a field application scientist (FAS) remotely or in your lab and helps you:
  • Get up and running quickly
  • Get connected to your local network and Cloud
  • Choose appropriate reagents and consumables to meet your needs
Content will vary depending on your level of knowledge and experience.
Service plans help protect your investment
  • Maximize system uptime with fast repair turnaround time from manufacturer-trained and certified field service engineers (FSEs)
  • Reduce overall repair costs with remote instrument diagnosis and resolution
  • Extend the life of your instrument and help keep it running at peak performance with annual maintenance service
During the annual maintenance, the FSE will update the software, if required; clean, lubricate, and align relevant parts and optical systems; and perform functional tests while documenting tasks, results, and findings.
Learn more about available financing options and programs
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Digital PCR
For Use With (Equipment)QuantStudio™ Absolute Q™ MAP16 Plate
Product LineQuantStudio
QuantitySystem + SmartStart + 1 yr ext warranty
Shipping ConditionRoom Temperature
Block FormatNon-interchangeable
Unit SizeEach
Contents & Storage
QuantStudio™ Absolute Q™ Digital PCR System, Desktop
Frequently asked questions (FAQs)
What is included in the QuantStudio Absolute Q Digital PCR System, desktop, extended warranty, Smart Start (Cat. No. A53267)?
The QuantStudio Absolute Q Digital PCR System, desktop, extended warranty, Smart Start (Cat. No. A53267) package includes the QuantStudio Absolute Digital PCR Instrument, Dell Tower and Dell Monitor, 1-year extended warranty, and SmartStart orientation. SmartStart orientation is a 1-day interactive course led by a Field Application Scientist (FAS) to help your team quickly become proficient using the software features and instrumentation.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
I finished running my experiment on the QuantStudio Absolute Q Digital PCR instrument but just realized that I had selected the wrong dyes during setup. Can I change the dyes that were used after the run?
If dye channels are turned off before the run, then data will not be collected for them as the run progresses. This means that changing the dyes after the run will not be possible.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
I ran my experiment using the QuantStudio Absolute Q Digital PCR System but did not actually use dyes in all the channels. Is there a way to only see those dye channels that were used?
Unused dye channels can be turned off after the run by going to the SETUP tab and clicking EDIT SETUP and then EDIT GROUPS. Here, the Analysis can be changed to “Not Used.” After clicking SAVE twice, the data will be reanalyzed and the unused dye channels will not be present in the analysis.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
I'm using the QuantStudio Absolute Q Digital PCR System and the software is saying that the concentration is NaN. What does that mean?
NaN stands for “not a number”. The software will display NaN instead of an actual number when it has detected some issue during the analysis of the array images.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
With the QuantStudio Absolute Q Digital PCR instrument, the run data didn't automatically transfer after the run completed. How can I get the data?
We would recommend restarting the software. If the run still doesn't transfer, we would recommend closing the software, rebooting the instrument, and then starting the software. If further assistance is needed, please contact Technical Support at techsupport@thermofisher.com.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Citations & References (20)
Citations & References
Abstract
Multidrug-resistant mutations to antiviral and antibody therapy in an immunocompromised patient infected with SARS-CoV-2.
Authors:Hirotsu Y, Kobayashi H, Kakizaki Y, Saito A, Tsutsui T, Kawaguchi M, Shimamura S, Hata K, Hanawa S, Toyama J, Miyashita Y, and Omata M
Journal:Med (New York, N.Y.)
PubMed ID:37683636
The circular RNA Ataxia Telangiectasia Mutated regulates oxidative stress in smooth muscle cells in expanding abdominal aortic aneurysms
Authors:Francesca Fasolo, Greg Winski, Zhaolong Li, Zhiyan Wu, Hanna Winter, Julia Ritzer, Nadiya Glukha, Joy Roy, Rebecka Hultgren, Jessica Pauli, Albert Busch, Nadja Sachs, Christoph Knappich, Hans-Henning Eckstein, Reinier A. Boon, Valentina Paloschi, and Lars Maegdefessel
Journal:Molecular Therapy Nucleic Acids
PubMed ID:
Monitoring of kinetics and exhaustion markers of circulating CAR-T cells as early predictive factors in patients with B-cell malignancies.
Authors:García-Calderón C, Sierro-Martínez B, García-Guerrero E, Sanoja-Flores L, Muñoz-García R, Ruiz-Maldonado V, Jimenez-Leon, Delgado-Serrano J, Molinos-Quintana A, Guijarro-Albaladejo B, Carrasco-Brocal I, Lucena J, García-Lozano J, Blázquez-Goñi C, Reguera-Ortega J, González-Escribano M, Reinoso-Segura M, Briones J, Pérez-Simón J, and Caballero-Velázque T
Journal:Frontiers in immunology
PubMed ID:37122702
Precision cancer monitoring using a novel, fully integrated, microfluidic array partitioning digital PCR platform.
Authors:Dueck M, Lin R, Zayac A, Gallagher S, Chao A, Jiang L, Datwani, S, Hung P, and Stieglitz E
Journal:Scientific reports
PubMed ID:31862911
The Digital MIQE Guidelines Update: Minimum Information for Publication of Quantitative Digital PCR Experiments for 2020.
Authors:Huggett JF
Journal:Clinical chemistry
PubMed ID:32746458