Product Image
Applied Biosystems™
GeneMapper™ Software v6.1, full installation
GeneMapper Software is a flexible genotyping software package that provides DNA sizing and quality allele calls for all Thermo FisherMás información
Have Questions?
Número de catálogoCantidad
A535471 licencia
Número de catálogo A53547
Precio (MXN)
-
Cantidad:
1 licencia
GeneMapper Software is a flexible genotyping software package that provides DNA sizing and quality allele calls for all Thermo Fisher Scientific electrophoresis-based genotyping systems. This software specializes in multi-application functionality, including amplified fragment length polymorphism (AFLP), loss of heterozygosity (LOH), microsatellite and SNP genotyping analysis. GeneMapper Software can help users increase data processing efficiency with remote auto-analysis and command line operation, and allows for multiuser, client-server deployment. The software uses Process Quality Values (PQVs) for automated identification that reduces data review time for high throughput genotyping. In addition, the security and audit features help users meet 21 CFR 11 requirements.
Features include:
• Installation on the data collection computer or a non-instrument computer
• Support of networked multiple clients when installed on a non-instrument computer
• An Oracle database with project-related data storage that ensures faster installation times and flexible archive capabilities
• Customizable project auto-saving frequency
• Defining of linearity range in the analysis methods for more reproducible, higher quality sizing of larger fragments (>800bps)
• Analysis of up to 8-dye data
GeneMapper Software has not undergone specific developmental validation for human identification applications. Human identification laboratories analyzing single-source or parentage samples that choose to use GeneMapper Software for data analysis should perform their own developmental validation studies.
Several users may access the database concurrently; however, because genotype analysis is data-intensive, more than five clients simultaneously accessing the database may slow the performance of GeneMapper Software.
The software is tested with the following specifications:
• Dell OptiPlex XE3 Windows™ 10 Enterprise LTSC operating system
• Dell 3551 Windows 10 Enterprise LTSC operating system
• CPU: Core i5-6440HQ/3.0 GHz and CoreTM i5-10400H CPU @ 2.60 GHz
• Memory: 16 GB
• Disk: Free space: 500 MB on the boot drive, 7 GB on the drive on which the GeneMapper Software is installed
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Introducción de datosCE Fragment Analysis (.fsa files)
LicenciaFull
Sistema operativoWindows 10
Cantidad1 licencia
Línea de productosGeneMapper
Software TypeCE Fragment Analysis
Unit SizeEach
Contenido y almacenamiento
The software is provided in the form of a USB stick.
Preguntas frecuentes
How can I set up auto-analysis for data in GeneMapper with the 3500 Data Collection Software version 3.x?
Beginning with 3500 Data Collection Software version 3, the 3500 Data Collection Software does not support automatic secondary analysis. This is also indicated in the software release notes. To perform secondary data analysis, you will need to manually import the .fsa file into the GeneMapper software. For optimal performance, we recommend that you install the GeneMapper Software on a stand-alone computer that is not part of the 3500 Genetic Analyzer system.
Find additional tips, troubleshooting help, and resources within our Fragment Analysis Software Support Center.
Can I install GeneMapper Software v6.1, full installation and GeneMapper Software v6.1, client installation on the same computer?
No, GeneMapper Full Software and GeneMapper Client Software must be installed on different computers.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
How many user accounts can be connected to the Oracle database embedded in GeneMapper Software v6.1, full installation at any time?
Five user accounts can be connected to the Oracle database embedded in GeneMapper Software v6.1, full installation at any time.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
We just upgraded to GeneMapper Software v6.1, and I'm wondering if my old data files will work on it.
GeneMapper Software v6.1 is compatible with the following files:
- 3130 series, Data Collection Software v3.0, 3.1, 4
- 3730 series, Data Collection Software v3.x, 4.x
- 3500 series, Data Collection Software v1.x, 2, 3.x
- SeqStudio Data Collection Software v1.x
- SeqStudio Flex Series Genetic Analyzer, Data Collection Software v1.x
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Where can I download the Release Notes for GeneMapper Software v6.1, full installation?
Please download this PDF (https://downloads.thermofisher.com/Instrument_Software/Genemapper_6.1_Full/100107864_GeneMapper-Full_v6.1_Release_Notes.pdf).
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.