CTS™ Viral Production Cells - FAQs
CTS™ Viral Production Cells - FAQs
View additional product information for CTS™ Viral Production Cells - FAQs (A53956)
3 product FAQs found
Are CTS Viral Production Cells 293T-derived?
No, CTS Viral Production Cells are HEK 293F suspension cells that have been adapted to CTS LV-MAX Production Medium. They do not express the SV40 large T antigen.
How do I bank CTS Viral Production Cells?
CTS Viral Production Cells can be frozen directly in cryogenic medium consisting of 90% CTS LV-MAX Production Medium plus 10% DMSO. Here is the protocol:
- Bank cells at passage 3 when cell density reaches ˜3.0-4.5 x 10E6 viable cells/mL at >95% viability.
- Centrifuge the cells at 100 x g (low speed) for 5 mins to pellet. Discard the spent medium.
- Prepare freezing medium: e.g., 10 mL of freezing medium is 9 mL of CTS LV-MAX Production Medium + 1 mL DMSO; mix well and sterilize by 0.2 µm filtration.
- Resuspend the cell pellet in the prepared freezing medium.
- Dilute the cells to a final density of 10 x10E6 viable cells/mL and aliquot 1 mL per cryovial.
- Freeze the cells at -80 degrees C for 1 day.
- Transfer frozen vials to liquid nitrogen for long-term storage.
Can I use CTS Viral Production Cells (Cat. No. A53956) with CTS AAV MAX products?
CTS Viral Production Cells 2.0 are specifically designed to work with CTS AAV MAX products to provide maximal adeno-associated virus (AAV) titers and optimal performance. CTS Viral Production Cells can be used but we don't recommend doing this as it will reduce system performance.