Can AccuCheck ERF Reference Particles (Cat. No. A55950) be used with fluorescence-activated cell sorters?
Yes, AccuCheck ERF Reference Particles (Cat. No. A55950) can be used with fluorescence-activated cell sorters as long as the cell sorter has the appropriate channel/filter(s).