Do you offer additional years of service for the iBlot 3 Western Transfer Device?
No, we do not provide service contracts or extended warranties for these devices. The warranty is limited to 1 year from the date of shipment.
Search
View additional product information for iBlot™ 3 Starter Kit, NC - FAQs (A56728, IB31001S)
1 product FAQs found