SureTect™ Escherichia coli STEC Identification PCR Assay
SureTect™ Escherichia coli STEC Identification PCR Assay
SureTect™ Escherichia coli STEC Identification PCR Assay
Thermo Scientific™

SureTect™ Escherichia coli STEC Identification PCR Assay

Rapidly detect multiple genetic targets for Shiga-toxin producing Escherichia coli O157:H7 and other STEC in food and environmental samples.

Have Questions?
Número do catálogo A56840
Preço (BRL)
-

The Thermo Scientific™ SureTect™ Escherichia coli STEC Identification PCR Assay is part of a complete workflow for the rapid detection of Escherichia coli O157:H7, stx, eae, and the Escherichia coli STEC serotypes (O26, O45, O103, O111, O121, O145) from food samples. The assay incorporates an optimized liquid lysis step, providing a quick and simple method for preparing the sample for rapid detection on the Applied Biosystems™ QuantStudio™ 5 Food Safety Real-Time PCR System and software.

Samples assessed as positive using the SureTect™ Escherichia coli O157:H7 and STEC Screening PCR Assay are further tested using this SureTect™ Escherichia coli STEC Identification PCR Assay. The assay has been AOAC-RI and AFNOR (ISO 16140-2:2016) validated on a range of food, including low and high sample sizes of raw meat (excluding poultry), vegetables, fruits, juices, and dairy samples, and it enables users to reliably detect and differentiate Shiga-toxin producing Escherichia coli O157:H7 and other STEC in as little as 10 hours.

The SureTect System is an optimized solution combining speed and performance in an easy-to-use, cost-effective platform—giving you results you can trust.

The new SureTect assay format incorporates color-coded plastics with orientation markings and breakable strip format, colored reagents and pierceable seals to enable an optimized workflow for maximum laboratory efficiency and productivity.

Simple

  • Easy-to-use with single enrichment step and pre-dispensed reagents
  • Target-specific colored assay plastics
  • Clearly oriented lysis and assay trays
  • Color indicator in lysis reagent reducing possibility for human error

Fast and Flexible

  • Option to automate the lysis and PCR sample preparation
  • Optimized workflow delivers on-screen results in as little as 10 hours
  • Maximum flexibility for operational efficiency
  • Run all assays simultaneously in the same run
  • Test up to 96 samples in each run
  • Choice of pierceable seals or caps
  • Detect multiple targets in the same run with universal PCR protocols across the range

Smart

  • Intuitive, user-friendly software delivers reliable results, with no subjective interpretation
  • Actionable data supports fast decisions in a complex product supply chain
  • IPC in every assay for complete confidence in results

The Thermo Scientific SureTect Escherichia coli STEC Identification PCR Assay Kit includes:

  • Escherichia coli STEC Identification PCR Tubes
  • PCR Caps

The lysate of screened positive samples is pipetted into the Escherichia coli STEC Identification PCR tubes to re-hydrate the pre-loaded PCR pellet which contains all the necessary components and reagents for PCR, including a probe, primers and DNA template for the internal amplification control (IAC). The PCR tubes are sealed, then loaded into the Applied Biosystems™ QuantStudio™ 5 Food Safety Real-Time PCR instrument using RapidFinder™ Analysis Software v3.0 or later. After completing the run, interpreted results are clearly displayed and can be reported, stored, printed or downloaded as required. The internal amplification control confirms correct assay conditions have occurred.

 Not all products are available for sale in all territories. Please inquire.

For Testing of Food and Environmental Samples Only.
Especificações
Product LineSureTect
Product TypeSureTect STEC Identification PCR
Target Organism ClassDetection of Escherichia coli O157:H7, stx, eae, and the Escherichia coli STEC serotypes (O26, O45, O103, O111, O121, O145)
For Use With (Application)Food and Beverage
Unit SizeEach