VetMAX™ Rotavirus A/B/C Reagents
Applied Biosystems™

VetMAX™ Rotavirus A/B/C Reagents

VetMAX Rotavirus A/B/C Reagents contain master mix, primer/probe mix, and nuclease-free water. The primer/probe mix contains oligos that enable single-wellRead more
Have Questions?
Catalog NumberQuantity
A57449100 reactions
Catalog number A57449
Price (TWD)
-
Quantity:
100 reactions

VetMAX Rotavirus A/B/C Reagents contain master mix, primer/probe mix, and nuclease-free water. The primer/probe mix contains oligos that enable single-well reverse-transcription real-time PCR (RT-qPCR) targeting porcine rotavirus groups A, B, and C, as well as Xeno RNA control (the internal positive control). The Rotavirus A/B/C master mix is a fast-cycling-capable, high-order multiplex-compatible master mix that has been designed to tolerate inhibition from challenging samples such as porcine feces.

Features of the VetMAX Rotavirus A/B/C Reagents include:
• Rapid and cost-effective method for amplification of target sequences
• Contains optimized fast-cycling, multiplex master mix
• Identifies and distinguishes rotavirus subtypes A, B, and C in a single well and from pools of up to five samples
• Fast run time of as little as 54 minutes

Rotaviruses (Reoviridae) are genetically diverse, non-enveloped, segmented, double-stranded RNA viruses that cause significant diarrheal disease in young pigs in commercial swine operations. Infection commonly causes villus atrophy resulting in dehydration, malabsorption, and wasting. Uncomplicated cases are typically accompanied by high morbidity but low mortality; however, co-infection with other enteric pathogens such as E. coli can lead to increased mortality rates.
 

For Research Use Only. Not for use in diagnostic procedures.
Specifications
DiseasePorcine Rotavirus
No. of Reactions100 reactions
Quantity100 reactions
Sample TypeFeces, intestinal samples, feedback samples
Shelf Life12 Months
Shipping ConditionShipped at -30°C to -10°C
Concentration2X (Master Mix), 10X (Primer/Probe Mix)
For Use With (Application)Real Time PCR (qPCR)
Unit SizeEach