TrueMark™ Enteric Bacterial Select Panel II Combo Kit
Applied Biosystems™

TrueMark™ Enteric Bacterial Select Panel II Combo Kit

The TrueMark Enteric Bacterial Select Panel II Combo Kit includes the TrueMark Enteric Bacterial Select Panel II and TrueMark InfectiousRead more
Have Questions?
Catalog NumberQuantity
A58076200 reactions
Catalog number A58076
Price (USD)
3,875.00
Each
Quantity:
200 reactions
Price (USD)
3,875.00
Each

The TrueMark Enteric Bacterial Select Panel II Combo Kit includes the TrueMark Enteric Bacterial Select Panel II and TrueMark Infectious Disease 1‐Step Multiplex Master Mix for simplified ordering. Together with the TrueMark Enteric Bacterial Select Amplification Control II (offered separately), the kit can be used to quickly evaluate and research up to 200 samples consisting of extracted nucleic acid from stool specimens.

The kit is recommended for use with the QuantStudio 5 Real-Time PCR System (with 96-well, 0.2-mL block). It includes primers and probe sets specific to the following targets:
• Vibrio (V. parahaemolyticus, V. vulnificus, and V. cholerae)
Yersinia enterocolitica
Plesiomonas shigelloides
Aeromonas hydrophila
Bacillus atrophaeus (used as a process control)

Features of the TrueMark Enteric Bacterial Select Panel II Combo Kit include:
• Detects and differentiates targets found in enteric bacterial infections
• Affordable and scalable—add to existing infrastructure to expand research menu while maintaining low operational costs
• Ready-to-use off-the-shelf solution—seamlessly integrate test into existing workflow

The TrueMark Enteric Bacterial Select Panel II Combo Kit includes the following components:
TrueMark Enteric Bacterial Select Panel II—multiplexed assay that contains primer and probe sets
TrueMark Infectious Disease 1‐Step Multiplex Master Mix (No ROX)—ready-to-use PCR mix, including polymerase, dNTPs, salt, and buffer

Use this kit with:
TrueMark Enteric Bacterial Select Amplification Control II—plasmid control containing target-specific templates

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For Research Use Only.
Specifications
No. of Reactions200 Reactions
Quantity200 reactions
Reaction FormatSingle tubes
Sample TypeStool
Detection MethodqPCR
For Use With (Application)Pathogen Detection, Bacterial Enteric Diagnostic Testing, Real Time PCR (qPCR), RT-PCR
FormLiquid
PCR MethodMultiplex qPCR, qPCR
Reaction Speed120 min.
Unit SizeEach