VetMAX™ African Swine Fever Virus I177L DIVA Controls
Applied Biosystems™

VetMAX™ African Swine Fever Virus I177L DIVA Controls

The VetMAX African Swine Fever Virus I177L DIVA Controls are synthetic positive controls used in the reflex qPCR assay for the rapid differentiation of infected animals vs animals vaccinated with the ASFV-GΔI177L recombinant vaccine (Cat. No. A59392).
Have Questions?
Catalog NumberQuantity
A59394500 reactions
Catalog number A59394
Price (USD)
-
Quantity:
500 reactions

The VetMAX African Swine Fever Virus I177L DIVA Controls are synthetic positive controls used in the reflex qPCR assay for the rapid differentiation of infected animals vs animals vaccinated with the ASFV-GΔI177L recombinant vaccine (Cat. No. A59392). This control is added to the assay to monitor for reaction failure or human error.

For Research Use Only.

Specifications
DiseaseAfrican Swine Fever
No. of Reactions500 Reactions
Quantity500 reactions
SpeciesSwine
Detection MethodqPCR
For Use With (Application)Pathogen Detection, Virus Detection
FormLiquid
GC-Rich PCR PerformanceMedium
Label or DyeVIC
PCR MethodqPCR
Reaction Speed30 to 50 min.
Unit SizeEach