What is the formulation of the Live Cell Imaging Solution (Cat. No. A59688DJ)?
The formulation of the Live Cell Imaging Solution (Cat. No. A59688DJ) is shown below:
140 mM NaCl
2.5 mM KCl
1.8 mM CaCl2
1.0 mM MgCl2
20 mM HEPES
pH 7.4
mOsm = 300
140 mM NaCl
2.5 mM KCl
1.8 mM CaCl2
1.0 mM MgCl2
20 mM HEPES
pH 7.4
mOsm = 300