Do you offer an alternative for SEAP Substrate Luminescence (Cat. No. A63880)?
We suggest the following products as an alternative to for SEAP Substrate Luminescence (Cat. No. A63880):
Phospha-Light SEAP Reporter Gene Assay System (Cat. Nos. T1015, T1016, T1017)
NovaBright Phospha-Light EXP Assay Kit for SEAP (Secreted Placental Alkaline Phosphatase) Reporter Gene Detection (Cat. Nos. N10577, N10578)
Both kits utilize the high-performance alkaline phosphatase substrate, CSPD, and an Emerald Enhancer.
Phospha-Light SEAP Reporter Gene Assay System (Cat. Nos. T1015, T1016, T1017)
NovaBright Phospha-Light EXP Assay Kit for SEAP (Secreted Placental Alkaline Phosphatase) Reporter Gene Detection (Cat. Nos. N10577, N10578)
Both kits utilize the high-performance alkaline phosphatase substrate, CSPD, and an Emerald Enhancer.