EquiPhi29™ DNA Amplification Kit
Thermo Scientific™

EquiPhi29™ DNA Amplification Kit

The Thermo Scientific™ EquiPhi29 DNA Amplification Kit includes all required components for fast, sensitive, and efficient isothermal DNA amplification, which includes rolling circle amplification (RCA) and multiple displacement amplification – whole genome amplification (MDA-WGA).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A65394500 Reactions
A65393100 Reactions
Catalog number A65394
Price (USD)
1,790.00
Each
Quantity:
500 Reactions
Price (USD)
1,790.00
Each

The Thermo Scientific™ EquiPhi29 DNA Amplification Kit includes all required components for fast, sensitive, and efficient isothermal DNA amplification, which includes rolling circle amplification (RCA) and multiple displacement amplification – whole genome amplification (MDA-WGA).

Features of the EquiPhi29 DNA Amplification Kit include:

  • Fast DNA amplification in less than two hours
  • Sensitivity from 1 fg of input DNA
  • Maximal yield of 17 μg amplified DNA
  • Compatibility with different types of sample input material (e.g., purified DNA, liquid media culture, agar plate colonies)
  • Compatibility with various downstream applications such as DNA sequencing (Sanger sequencing, next-generation sequencing), digestion with restriction enzymes, cell-free DNA enrichment, and cell-free protein expression
  • All required components for DNA amplification in RCA or MDA-WGA reactions

The kit contains EquPhi29 DNA Polymerase, a proprietary phi29 DNA polymerase mutant that possesses strong strand displacement activity and enables high thermostability, reaction speed, and product yield and low amplification bias. Also included is an optimized reaction buffer, exo-resistant random primer, pyrophosphatase, and dNTPs. The addition of pyrophosphatase to the reaction mixture enhances DNA amplification and facilitates sample handling.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
FormatKit
Optimal Reaction Temperature42°C
Reaction TimeUp to 2 hours
Shipping ConditionDry Ice
For Use With (Application)RCA, MDA-WGA
FormLiquid
PolymeraseEquiPhi29 DNA Polymerase
Product TypeDNA Amplification Kit
Quantity500 Reactions
Research CategoryNucleic Acid Analysis, In Vitro Transcription, Cell-free Protein Expression, In Situ Sequencing, SNP Detection
Unit SizeEach
Contents & Storage

• 5 x 100 μL EquiPhi29 DNA Polymerase (10 U/μL)
• 2 x 1 mL EquiPhi29 DNA Polymerase Reaction Buffer (10X)
• 0.25 mL DTT (0.1 M)
• 5 x 200 μL Exo-Resistant Random Primer (500 μM)
• 1 mL dNTP Mix (10 mM each)
• 5 x 100 μL Pyrophosphatase, inorganic (0.1 U/μL)

Store at -15 to -25°C.

Frequently asked questions (FAQs)

What is the error rate of EquiPhi29 DNA Polymerase?

The error rate of EquiPhi29 DNA Polymerase is 6 x 10-6.
The error rate of EquiPhi29 DNA Polymerase was measured according to the method described in literature:
Mielinis, P., Sukackaitė, R., Serapinaitė, A., Samoilovas, F., Alzbutas, G., Matjošaitis, K., & Lubys, A. (2021). MUA-based molecular indexing for rare mutation detection by Next-Generation sequencing. Journal of Molecular Biology, 433(19), 167209. https://doi.org/10.1016/j.jmb.2021.167209

Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.

Can I use EquiPhi29 DNA Polymerase to incorporate 5-methyl-dCTP?

Thermo Scientific EquiPhi29 DNA Polymerase is a proprietary mutant phi29 DNA Polymerase developed through in vitro protein evolution. EquiPhi29 DNA Polymerase as well as phi29 DNA Polymerase should be able to incorporate 5-methyl-dCTP nucleotides and other modified nucleotides.

Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.

When using EquiPhi29 DNA Polymerase, do I need to purify amplified DNA products before downstream applications?

Cleaning of the amplified product is not required prior to several downstream methods (e.g., debranching, digestion with restriction endonucleases, Sanger sequencing); the dilution of amplified product is sufficient. If the clean-up procedure is needed, we recommend using an affinity-based spin-column or magnetic bead-based purification method.

Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.

What is the minimal recommended time for amplification with EquiPhi29 DNA Polymerase?

The optimal reaction time for DNA amplification with EquiPhi29 DNA Polymerase is 2 hours. For samples with ≥1 pg of DNA input, DNA amplification time can be shortened to 1 hour if maximizing product yield is not essential.

Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.

Can liquid media culture or colonies be used as a starting material for amplification with EquiPhi29 DNA Polymerase?

Yes. EquiPhi29 DNA Polymerase can work with different types of sample input material such as purified DNA, liquid media culture, agar plate colonies, etc.

Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.