CTS™ StemPro™-34 SFM XF Medium
Gibco™
CTS™ StemPro™-34 SFM XF Medium
Gibco CTS StemPro-34 SF XenoFree Medium is a flexible, optimized medium for hematopoietic stem and progenitor cell applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityProduct Type
A66369022 LBPC Kit
A6636901500 mLBottle Kit
Catalog number A6636902
Price (USD)
2,560.00
Each
Quantity:
2 L
Product Type:
BPC Kit
Price (USD)
2,560.00
Each
Gibco CTS StemPro-34 SF XenoFree Medium is a highly effective medium designed for the growth and development of human hematopoietic cells in culture, including hematopoietic stem cells (HSCs) and progenitor cells sourced from bone marrow, neonatal cord blood, and peripheral blood.
Key benefits for cell and gene therapy applications
  • Serum-free and xeno-free—formulated without serum or non-human derived components to comply with global regulatory standards for ancillary materials in cell therapy manufacturing; formulated with L-glutamine and without phenol red
  • Supports cell differentiation—facilitates differentiation of induced pluripotent stem cells (iPSCs) to HSCs and further differentiation into immune cells (NK cells, T-cells, B-cells, macrophages, etc.)
Regulatory compliance and quality assurance
  • cGMP manufactured—produced in accordance with 21 CFR part 820 and ISO 9001 and ISO 13485 standards to help ensure the highest quality and safety
  • USP <1043> compliant—designed to meet USP <1043> standards for ancillary materials used in cell, gene, and tissue-engineered products
Seamless integration with closed system workflows
CTS StemPro-34 SF XenoFree Medium is available in both 500-mL bottle and 2-liter bioprocessing container (BPC) formats, supporting processes through cell therapy development and manufacturing. The BPC chamber is constructed from Aegis 5-14 film and includes sterile, weldable DEHP-free PVC tubing with Luer lock and C-Flex tubing with MPC quick connector. This helps provide versatile connection options, helping ensure maximum sterility and efficiency in your manufacturing process. The 2-L kit format includes both supplement and basal components in BPCs for closed system workflows.
Comprehensive documentation for regulatory support
To facilitate your regulatory submissions, CTS StemPro-34 SF XenoFree Medium comes with an extensive documentation package, including certificates of analysis (COA), access to drug master files (DMFs), and/or regulatory support files (RSFs), and certificates of origin (COO) upon request.
Choose CTS StemPro-34 SF XenoFree Medium for a robust, regulatory-compliant solution that supports the critical processes in cell and gene therapy manufacturing, helping ensure the highest standards of quality and safety.
For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.
Specifications
Product TypeBPC Kit
Quantity2 L
ClassificationXeno-free, Serum-free
Culture TypeStem Cell (Human, iPS - Induced Pluripotent Stem, Embryonic)
FormLiquid
Serum LevelSerum-free
SterilitySterile-filtered
Sterilization MethodSterile-filtered
With AdditivesGlutamine
Without AdditivesNo Phenol Red
Unit SizeEach
Frequently asked questions (FAQs)
What is CTS?
The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.
Once thawed, can CTS StemPro-34 Supplement in bags be refrozen?
Once thawed, CTS StemPro-34 Supplement should not be refrozen in the bag. If necessary, the supplement can be removed from the bag to a secondary container and refrozen. Avoid additional freeze/thaw cycles.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How long can complete CTS StemPro-34 SFM XF medium (basal medium plus supplement) be stored at 4 degrees C?
Complete CTS StemPro-34 SFM XF medium can be stored for 4 weeks at 4 degrees C, however, we recommend adding growth factors or cytokines just prior to use.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How do I thaw and mix CTS StemPro-34 Supplement?
CTS StemPro-34 Supplement can be thawed at room temperature (RT) for ~1 hour or at 4 degrees C overnight. Do not thaw the frozen supplement at 37 degrees C. Once completely thawed, the supplement should be allowed to equilibrate to RT for at least 10 min, but not longer than 3 hr, before gently mixing the supplement by inverting the vial 6-8 times until the supplement is completely homogenous.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How should I store the components of the CTS StemPro-34 SFM XF kit?
CTS StemPro-34 Basal Medium should be stored at 2 to 8 degrees C, protected from light. CTS StemPro-34 Supplement should be stored at -20 to -5 degrees C, protected from light.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.