CTS™ StemPro™-34 SFM XF Medium, BPC, 2 L - FAQs
CTS™ StemPro™-34 SFM XF Medium, BPC, 2 L - FAQs
View additional product information for CTS™ StemPro™-34 SFM XF Medium - FAQs (A6636901, A6636902)
8 product FAQs found
What is CTS?
The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.
Once thawed, can CTS StemPro-34 Supplement in bags be refrozen?
Once thawed, CTS StemPro-34 Supplement should not be refrozen in the bag. If necessary, the supplement can be removed from the bag to a secondary container and refrozen. Avoid additional freeze/thaw cycles.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How long can complete CTS StemPro-34 SFM XF medium (basal medium plus supplement) be stored at 4 degrees C?
Complete CTS StemPro-34 SFM XF medium can be stored for 4 weeks at 4 degrees C, however, we recommend adding growth factors or cytokines just prior to use.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How do I thaw and mix CTS StemPro-34 Supplement?
CTS StemPro-34 Supplement can be thawed at room temperature (RT) for ~1 hour or at 4 degrees C overnight. Do not thaw the frozen supplement at 37 degrees C. Once completely thawed, the supplement should be allowed to equilibrate to RT for at least 10 min, but not longer than 3 hr, before gently mixing the supplement by inverting the vial 6-8 times until the supplement is completely homogenous.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How should I store the components of the CTS StemPro-34 SFM XF kit?
CTS StemPro-34 Basal Medium should be stored at 2 to 8 degrees C, protected from light. CTS StemPro-34 Supplement should be stored at -20 to -5 degrees C, protected from light.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Does CTS StemPro-34 SFM XF contain any added growth factors or cytokines?
CTS StemPro‑34 SFM XF is manufactured without cytokines or growth factors, to allow the researcher flexibility for the intended application. Please refer to the user guide for details.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What types of cells have been tested with CTS StemPro-34 SFM XF?
CTS StemPro-34 SFM XF can support growth of human hematopoietic stem and progenitor cells in culture, including CD34+ cells from bone marrow, neonatal cord blood, and peripheral blood, when supplemented with appropriate growth factors or cytokines. In addition, CTS StemPro-34 SFM XF can support differentiation of human induced pluripotent stem cells (iPSCs) to iPSC-derived HSPCs, when supplemented with appropriate growth factors, cytokines and other additions. A suggested protocol is available here.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What are the main differences between StemPro-34 SFM and CTS StemPro-34 SFM XF?
CTS Stempro-34 SFM XF is for research use or manufacturing of cell-, gene-, or tissue-based products. It is not intended for direct administration into humans or animals. It is serum-free and xeno-free, contains L-Glutamine and does not contain phenol red. For a detailed description of CTS StemPro-34 SFM XF, please refer to the Product Overview section on the product page.
StemPro-34 SFM is designed for research use only, it is serum-free, contains phenol red, and requires addition of a source of L-Glutamine.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.