Tubes, 0.2 mL, flat cap - FAQs
Tubes, 0.2 mL, flat cap - FAQs
View additional product information for Tubes, 0.2 mL, flat cap - FAQs (AB0620)
4 product FAQs found
Are Thermo Scientific ABgene plastics endotoxin-free?
They are not certified endotoxin-free. However, their entire production process, from molding to final packaging, is carried out in a Class 100,000 clean room under ISO 9001 guidelines. All Thermo Scientific PCR plastics are certified free from RNase, DNase and human DNA.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Are Thermo Scientific ABgene plastics sterile?
Thermo Scientific Abgene plastics are not typically sterilized as their entire production process, from molding to final packaging, is carried out in a Class 100,000 clean room under ISO 9001 guidelines. All Thermo Scientific PCR plastics are certified free from RNase, DNase and human DNA. In contrast, during typical non-clean room production, plastics are exposed to many contaminants including dust, bacterial cells, and DNA. The plastics are then sterilized to kill bacteria and inactivate RNases and DNases, but sterilization does not remove dust or DNA contamination. The dust particles left behind can inhibit PCR, and the damaged DNA fragments can still act as templates, leading to non-specific amplification.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Are the Tubes, 0.2 mL, flat cap (Cat. No. AB0620) suitable for storage at -80°C?
No, they are not. Due to the design of the tubes for PCR reactions they are not suitable for storage below -20°C, and therefore, we do not at all recommend to store them at -80°C.
Find additional tips, troubleshooting help, and resources within our PCR Thermal Cyclers Support Center.
Are the Tubes, 0.2 mL, flat cap (Cat. No. AB0620) also available in a strip format, i.e., as multiple tubes attached to each other?
Yes. Please see the following catalog numbers: AB1182, AB1114, and A30588.