A more sustainable equivalent is available.
Domed and Flat PCR Cap Strips
Thermo Scientific™
Domed and Flat PCR Cap Strips
The Thermo Scientific Flat PCR Cap ,Strip of 8 are used for PCR applications and work with the Thermo ScientificRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityColor
AB0784250/Cs.Clear
AB0783120/Cs.Clear
Catalog number AB0784
Price (USD)
101.65
Online exclusive
121.00
Save 19.35
Each
Quantity:
250/Cs.
Color:
Clear
Request bulk or custom format
Price (USD)
101.65
Online exclusive
121.00
Save 19.35
Each
The Thermo Scientific Flat PCR Cap ,Strip of 8 are used for PCR applications and work with the Thermo Scientific 96-well PCR plates and strip tubes. These cap strips are available in either 8- or 12-cap strips. The Flat PCR Cap Strips signature flat cap allows you to label each sample.
Features of the Thermo Scientific Flat PCR Cap Strip
• 8- and 12-cap strips
• Suitable for use with 96-well PCR plates as well as standard and low profile strips
• Flat surface of each cap facilitates sample labeling
Related Products
Flat PCR Caps, strips of 8
Flat PCR Caps, strips of 12
Specifications
FormatStrips of 8 Flat Caps
Product TypePCR Plate Cap Strip
Quantity250/Cs.
Certified RNase/DNase-freeYes
ColorClear
MaterialPolypropylene
No. of Wells8
Plate Compatibility96-well Plate
Unit SizeEach
Frequently asked questions (FAQs)
Which caps are Thermo-Fast PCR Plate, 24-well (Cat. No. AB0624) compatible with?
This plate is compatible with all caps e.g. Flat PCR Caps, strips of 8 (Cat. No. AB0784) and Domed PCR Caps, strips of 8 (Cat. No. AB0265).
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.