Which caps are Thermo-Fast PCR Plate, 24-well (Cat. No. AB0624) compatible with?
This plate is compatible with all caps e.g. Flat PCR Caps, strips of 8 (Cat. No. AB0784) and Domed PCR Caps, strips of 8 (Cat. No. AB0265).