Extensor Long PCR ReddyMix Master Mix, with Buffer 1 - FAQs
Extensor Long PCR ReddyMix Master Mix, with Buffer 1 - FAQs
View additional product information for Extensor Long PCR ReddyMix Master Mix, with Buffer 1 - FAQs (AB0794B)
1 product FAQs found
How important is the quality of my DNA template in long PCR?
Template preparation becomes particularly important when performing longer amplifications (>15 kb). Therefore, it is recommended to check the length of the DNA by agarose gel electrophoresis.