Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
PCR Plate, 96-well, segmented, semi-skirted, green
Thermo Scientific™

PCR Plate, 96-well, segmented, semi-skirted, green

The Thermo Scientific 96-Well Semi-Skirted Plates, Green, Segmented provide superior rigidity and provides space for barcoding. The patented segmented designRead more
Have Questions?
Catalog NumberQuantity
AB0900G25 plates
Catalog number AB0900G
Price (USD)
171.00
Each
Quantity:
25 plates
Request bulk or custom format
Price (USD)
171.00
Each
The Thermo Scientific 96-Well Semi-Skirted Plates, Green, Segmented provide superior rigidity and provides space for barcoding. The patented segmented design of this plate allows it to be cut into 24 and 48 well sections.

Features of The Thermo Scientific 96-Well Semi-Skirted Plates, Segmented:

• Patented segmented plate design—allows plates to be cut into 24 and 48 well sections
• Semi skirt—adds rigidity and allows for labeling or barcoding
• Raised rim—around each well enables secure sealing and safeguards against evaporation
• 0.3 mL maximum well volume—when used with adhesive and heat seals
• Uniform, thin well walls—deliver maximum and consistent heat transfer
• White plates—optimized for qPCR by increasing sensitivity and reducing variability in fluorescence detection
• Cut corner: H1

Applications

• PCRRelated Products
PCR Plate, 96-well, segmented, semi-skirted
PCR Plate, 96-well, segmented, semi-skirted, blue
PCR Plate, 96-well, segmented, semi-skirted, green
PCR Plate, 96-well, segmented, semi-skirted, black
PCR Plate, 96-well, segmented, semi-skirted, purple
PCR Plate, 96-well, segmented, semi-skirted, red
PCR Plate, 96-well, segmented, semi-skirted, white
PCR Plate, 96-well, segmented, semi-skirted, yellow
PCR Plate, 96-well, segmented, semi-skirted, barcoded
PCR Plate, 96-well, segmented, semi-skirted, white, barcoded
Specifications
Certifications/ComplianceCertified RNase/DNase-free
For Use With (Equipment)Real-Time PCR Systems or Thermal Cyclers
Packaging25 Plate
Product TypePCR Microplate
Quantity25 plates
Reaction SpeedStandard
Skirt StyleSemi
Volume (Metric) Well0.3 mL
Volume (Metric) Working0.2 mL
BarcodeNo Barcode
ColorGreen
MaterialPolypropylene
No. of Wells96
Unit SizeEach