Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
Tubes and Flat Caps, strips of 12
Thermo Scientific™

Tubes and Flat Caps, strips of 12

Thermo Scientific Tubes and Flat Caps, Strips of 12 are designed for PCR applications, and are suitable for 0.2 mLRead more
Have Questions?
Catalog NumberQuantity
AB111480 strips
Catalog number AB1114
Price (USD)
210.00
Each
Quantity:
80 strips
Request bulk or custom format
Price (USD)
210.00
Each
Thermo Scientific Tubes and Flat Caps, Strips of 12 are designed for PCR applications, and are suitable for 0.2 mL thermal cycler blocks and 96-well V-bottom thermal cyclers. The 8-strip tubes can be divided into individual tubes.

Features of Thermo Scientific Tubes and Flat Caps strips:

• Suitable for 0.2 mL thermal cycler blocks
• 0.25 mL maximum tube capacity when capped
• Ideal for use in 96-well V-bottom thermal cyclers
• 10 packs of 8

Applications

• PCR

Related Products
Tubes and Domed Caps, strips of 12
Tubes and Flat Caps, strips of 8
Specifications
Certifications/ComplianceCertified DNA/RNase Free
For Use With (Equipment)Thermal Cyclers
Format80 Strip Tubes and Caps
No. of Wells96
Quantity80 strips
Reaction SpeedStandard
Volume (Metric) Working0.2 mL
Closure TypeFlat Cap
ColorNatural
MaterialPolypropylene
Product Type12-Strip Tube
SterilityNon-sterile
Volume (Metric)0.25 mL
Unit SizeEach

Frequently asked questions (FAQs)

Are the Tubes, 0.2 mL, flat cap (Cat. No. AB0620) also available in a strip format, i.e., as multiple tubes attached to each other?

Yes. Please see the following catalog numbers: AB1182, AB1114, and A30588.