Are the Tubes, 0.2 mL, flat cap (Cat. No. AB0620) also available in a strip format, i.e., as multiple tubes attached to each other?
Yes. Please see the following catalog numbers: AB1182, AB1114, and A30588.
Search
View additional product information for Tubes and Flat Caps, strips of 12 - FAQs (AB1114)
1 product FAQs found